missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-(Boc-amino)cyclopentanecarboxylic acid, 98%, Thermo Scientific Chemicals
$74.34 - $74.34
Chemical Identifiers
CAS | 35264-09-6 |
---|---|
Molecular Formula | C11H19NO4 |
Molecular Weight (g/mol) | 229.276 |
MDL Number | MFCD01076126 |
InChI Key | YBZCSKVLXBOFSL-UHFFFAOYSA-N |
Synonym | boc-cycloleucine, 1-boc-amino cyclopentanecarboxylic acid, 1-tert-butoxycarbonylamino cyclopentanecarboxylic acid, 1-n-boc-aminocyclopentanecarboxylic acid, boc-1-aminocyclopentane-1-carboxylic acid, n-boc-cycloleucine, 1-tert-butoxycarbonyl amino cyclopentanecarboxylic acid, 1-tert-butoxycarbonylamino-cyclopentanecarboxylic acid, 1-tert-butoxy carbonyl amino cyclopentane-1-carboxylic acid |
PubChem CID | 2734645 |
IUPAC Name | 1-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopentane-1-carboxylic acid |
SMILES | CC(C)(C)OC(=O)NC1(CCCC1)C(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1948303
|
Thermo Scientific Chemicals
L1948303 |
1 g |
Each for $74.34
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
35264-09-6 | |
229.276 | |
YBZCSKVLXBOFSL-UHFFFAOYSA-N | |
2734645 | |
CC(C)(C)OC(=O)NC1(CCCC1)C(=O)O |
C11H19NO4 | |
MFCD01076126 | |
boc-cycloleucine, 1-boc-amino cyclopentanecarboxylic acid, 1-tert-butoxycarbonylamino cyclopentanecarboxylic acid, 1-n-boc-aminocyclopentanecarboxylic acid, boc-1-aminocyclopentane-1-carboxylic acid, n-boc-cycloleucine, 1-tert-butoxycarbonyl amino cyclopentanecarboxylic acid, 1-tert-butoxycarbonylamino-cyclopentanecarboxylic acid, 1-tert-butoxy carbonyl amino cyclopentane-1-carboxylic acid | |
1-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopentane-1-carboxylic acid |
Specifications
131°C to 134°C | |
C11H19NO4 | |
2734412 | |
YBZCSKVLXBOFSL-UHFFFAOYSA-N | |
1-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopentane-1-carboxylic acid | |
2734645 | |
98% | |
1-(Boc-amino)cyclopentanecarboxylic acid |
35264-09-6 | |
MFCD01076126 | |
boc-cycloleucine, 1-boc-amino cyclopentanecarboxylic acid, 1-tert-butoxycarbonylamino cyclopentanecarboxylic acid, 1-n-boc-aminocyclopentanecarboxylic acid, boc-1-aminocyclopentane-1-carboxylic acid, n-boc-cycloleucine, 1-tert-butoxycarbonyl amino cyclopentanecarboxylic acid, 1-tert-butoxycarbonylamino-cyclopentanecarboxylic acid, 1-tert-butoxy carbonyl amino cyclopentane-1-carboxylic acid | |
CC(C)(C)OC(=O)NC1(CCCC1)C(=O)O | |
229.276 | |
229.28 | |
1 g |
Safety and Handling
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only