missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromo-2-butanone, 90%, stabilized, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 222450050

 View more versions of this product

Catalog No. AC222450050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

1-Bromo-2-butanone
816-40-0
100.0
105°C (150.0mmHg)
Authentic
Glass bottle
1.4650 to 1.469
MFCD00000207
01,67
1-bromo-2-butanone, 2-butanone, 1-bromo, 1-bromobutanone, bromomethyl ethyl ketone, bromobutanone, bromobutan-2-one, l-bromo-2-butanone, acmc-20aoxd, 1-bromo-butan-2-one
CCC(=O)CBr
151
13156
90%
Stabilized
88.0
1.4790g/mL
68°C
88% min. (GC)
C4H7BrO
C2H5COCH2Br
5g
1.479
CCXQVBSQUQCEEO-UHFFFAOYSA-N
1-bromobutan-2-one
approx. 0.5% CaCO3
151

Chemical Identifiers

816-40-0
151
CCXQVBSQUQCEEO-UHFFFAOYSA-N
13156
CCC(=O)CBr
C4H7BrO
MFCD00000207
1-bromo-2-butanone, 2-butanone, 1-bromo, 1-bromobutanone, bromomethyl ethyl ketone, bromobutanone, bromobutan-2-one, l-bromo-2-butanone, acmc-20aoxd, 1-bromo-butan-2-one
1-bromobutan-2-one
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor

GHS Signal Word: Warning

EINECSNumber : 212-431-3

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only