missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromo-2-butanone, 90%, stabilized, Thermo Scientific™

Chemical Identifiers

CAS 816-40-0
Molecular Formula C4H7BrO
Molecular Weight (g/mol) 151
MDL Number MFCD00000207
InChI Key CCXQVBSQUQCEEO-UHFFFAOYSA-N
Synonym 1-bromo-2-butanone, 2-butanone, 1-bromo, 1-bromobutanone, bromomethyl ethyl ketone, bromobutanone, bromobutan-2-one, l-bromo-2-butanone, acmc-20aoxd, 1-bromo-butan-2-one
PubChem CID 13156
IUPAC Name 1-bromobutan-2-one
SMILES CCC(=O)CBr
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC222450010
SDS View Documents
Thermo Scientific Chemicals
222450010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC222450050
SDS View Documents
Thermo Scientific Chemicals
222450050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC222450250
SDS View Documents
Thermo Scientific Chemicals
222450250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

816-40-0
151
CCXQVBSQUQCEEO-UHFFFAOYSA-N
13156
CCC(=O)CBr
C4H7BrO
MFCD00000207
1-bromo-2-butanone, 2-butanone, 1-bromo, 1-bromobutanone, bromomethyl ethyl ketone, bromobutanone, bromobutan-2-one, l-bromo-2-butanone, acmc-20aoxd, 1-bromo-butan-2-one
1-bromobutan-2-one

Specifications

816-40-0
100.0
105°C (150.0mmHg)
Authentic
Glass bottle
1.4650 to 1.469
MFCD00000207
01,67
1-bromo-2-butanone, 2-butanone, 1-bromo, 1-bromobutanone, bromomethyl ethyl ketone, bromobutanone, bromobutan-2-one, l-bromo-2-butanone, acmc-20aoxd, 1-bromo-butan-2-one
CCC(=O)CBr
151
13156
90%
88.0
1.4790g/mL
68°C
88% min. (GC)
C4H7BrO
C2H5COCH2Br
1g
1.479
CCXQVBSQUQCEEO-UHFFFAOYSA-N
1-bromobutan-2-one
approx. 0.5% CaCO3
151
1-Bromo-2-butanone, Stabilized
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor

GHS Signal Word: Warning

EINECSNumber : 212-431-3

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only