Learn More
1-Bromo-2-butanone, 90%, stabilized, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 222450250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| 1-Bromo-2-butanone | |
| 816-40-0 | |
| 100.0 | |
| 105°C (150.0mmHg) | |
| Authentic | |
| Glass bottle | |
| 1.4650 to 1.469 | |
| MFCD00000207 | |
| 01,67 | |
| 1-bromo-2-butanone, 2-butanone, 1-bromo, 1-bromobutanone, bromomethyl ethyl ketone, bromobutanone, bromobutan-2-one, l-bromo-2-butanone, acmc-20aoxd, 1-bromo-butan-2-one | |
| CCC(=O)CBr | |
| 151 | |
| 13156 | |
| 90% |
| Stabilized | |
| 88.0 | |
| 1.4790g/mL | |
| 68°C | |
| 88% min. (GC) | |
| C4H7BrO | |
| C2H5COCH2Br | |
| 25g | |
| 1.479 | |
| CCXQVBSQUQCEEO-UHFFFAOYSA-N | |
| 1-bromobutan-2-one | |
| approx. 0.5% CaCO3 | |
| 151 |
Chemical Identifiers
| 816-40-0 | |
| 151 | |
| CCXQVBSQUQCEEO-UHFFFAOYSA-N | |
| 13156 | |
| CCC(=O)CBr |
| C4H7BrO | |
| MFCD00000207 | |
| 1-bromo-2-butanone, 2-butanone, 1-bromo, 1-bromobutanone, bromomethyl ethyl ketone, bromobutanone, bromobutan-2-one, l-bromo-2-butanone, acmc-20aoxd, 1-bromo-butan-2-one | |
| 1-bromobutan-2-one |
Safety and Handling
GHS H Statement
Harmful if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor
GHS Signal Word: Warning
EINECSNumber : 212-431-3
RUO â Research Use Only