missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromo-2-chloroethane, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 214181000

 View more versions of this product

Catalog No. AC214181000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Bromo-2-chloroethane
97.5
-17.0°C
1.7300g/mL
Authentic
Glass bottle
1.4900 to 1.4920
MFCD00000962
1.73
Solubility in water: 7g/L (20°C). Other solubilities: soluble in alcohol and ether
C(CBr)Cl
143.41
CHEBI:19032
98%
107-04-0
100.0
Colorless to Yellow
106.0°C to 107.0°C
97.5% min. (GC)
C2H4BrCl
ClCH2CH2Br
100 mL
ethylene chlorobromide, 2-chloroethyl bromide, 1-chloro-2-bromoethane, 2-bromoethyl chloride, ethane, 1-bromo-2-chloro, 2-bromo-1-chloroethane, s-chlorobromoethane, 1,2-bromochloroethane, beta-chloroethyl bromide, sym-chlorobromoethane
IBYHHJPAARCAIE-UHFFFAOYSA-N
1-bromo-2-chloroethane
7849
143.41
Liquid

Chemical Identifiers

107-04-0
143.41
IBYHHJPAARCAIE-UHFFFAOYSA-N
7849
1-bromo-2-chloroethane
C2H4BrCl
MFCD00000962
ethylene chlorobromide, 2-chloroethyl bromide, 1-chloro-2-bromoethane, 2-bromoethyl chloride, ethane, 1-bromo-2-chloro, 2-bromo-1-chloroethane, s-chlorobromoethane, 1,2-bromochloroethane, beta-chloroethyl bromide, sym-chlorobromoethane
CHEBI:19032
C(CBr)Cl
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Toxic if inhaled.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove to fresh air

GHS Signal Word: Danger

EINECSNumber : 203-456-

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only