Learn More
1-Bromo-2-chloroethane, 98%, Thermo Scientific Chemicals
$120.25 - $120.25
Chemical Identifiers
CAS | 107-04-0 |
---|---|
Molecular Formula | C2H4BrCl |
Molecular Weight (g/mol) | 143.41 |
MDL Number | MFCD00000962 |
InChI Key | IBYHHJPAARCAIE-UHFFFAOYSA-N |
Synonym | ethylene chlorobromide, 2-chloroethyl bromide, 1-chloro-2-bromoethane, 2-bromoethyl chloride, ethane, 1-bromo-2-chloro, 2-bromo-1-chloroethane, s-chlorobromoethane, 1,2-bromochloroethane, beta-chloroethyl bromide, sym-chlorobromoethane |
PubChem CID | 7849 |
ChEBI | CHEBI:19032 |
IUPAC Name | 1-bromo-2-chloroethane |
SMILES | C(CBr)Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC214181000
|
Thermo Scientific Chemicals
214181000 |
100 mL | Glass bottle |
Each for $120.25
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
107-04-0 | |
143.41 | |
IBYHHJPAARCAIE-UHFFFAOYSA-N | |
7849 | |
1-bromo-2-chloroethane |
C2H4BrCl | |
MFCD00000962 | |
ethylene chlorobromide, 2-chloroethyl bromide, 1-chloro-2-bromoethane, 2-bromoethyl chloride, ethane, 1-bromo-2-chloro, 2-bromo-1-chloroethane, s-chlorobromoethane, 1,2-bromochloroethane, beta-chloroethyl bromide, sym-chlorobromoethane | |
CHEBI:19032 | |
C(CBr)Cl |
Specifications
107-04-0 | |
100.0 | |
Colorless to Yellow | |
106.0°C to 107.0°C | |
97.5% min. (GC) | |
C2H4BrCl | |
ClCH2CH2Br | |
100 mL | |
ethylene chlorobromide, 2-chloroethyl bromide, 1-chloro-2-bromoethane, 2-bromoethyl chloride, ethane, 1-bromo-2-chloro, 2-bromo-1-chloroethane, s-chlorobromoethane, 1,2-bromochloroethane, beta-chloroethyl bromide, sym-chlorobromoethane | |
IBYHHJPAARCAIE-UHFFFAOYSA-N | |
1-bromo-2-chloroethane | |
7849 | |
143.41 | |
Liquid |
97.5 | |
-17.0°C | |
1.7300g/mL | |
Authentic | |
Glass bottle | |
1.4900 to 1.4920 | |
MFCD00000962 | |
1.73 | |
Solubility in water: 7g/L (20°C). Other solubilities: soluble in alcohol and ether | |
C(CBr)Cl | |
143.41 | |
CHEBI:19032 | |
98% | |
1-Bromo-2-chloroethane |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Toxic if inhaled.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove to fresh air
GHS Signal Word: Danger
EINECSNumber : 203-456-
RUO – Research Use Only