missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromo-3-fluoro-5-nitrobenzene, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H61192MD

 View more versions of this product

Catalog No. AAH61192MD


Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

1-Bromo-3-fluoro-5-nitrobenzene
Yellow
MFCD09955454
UN2810
SWXVEPMSQBEVRH-UHFFFAOYSA-N
1-bromo-3-fluoro-5-nitrobenzene
15020157
98%
7087-65-2
C6H3BrFNO2
250 mg
3-bromo-5-fluoronitrobenzene, 1-nitro-3-fluoro-5-bromobenzene, benzene, 1-bromo-3-fluoro-5-nitro, pubchem17569, acmc-209ogb, 3-fluoro-5-nitrobromobenzene, 5-bromo-3-fluoro-1-nitrobenzene
C1=C(C=C(C=C1F)Br)[N+](=O)[O-]
219.997
220
Liquid

Chemical Identifiers

7087-65-2
219.997
SWXVEPMSQBEVRH-UHFFFAOYSA-N
15020157
C1=C(C=C(C=C1F)Br)[N+](=O)[O-]
C6H3BrFNO2
MFCD09955454
3-bromo-5-fluoronitrobenzene, 1-nitro-3-fluoro-5-bromobenzene, benzene, 1-bromo-3-fluoro-5-nitro, pubchem17569, acmc-209ogb, 3-fluoro-5-nitrobromobenzene, 5-bromo-3-fluoro-1-nitrobenzene
1-bromo-3-fluoro-5-nitrobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions