Learn More
1-Bromo-3-fluoro-5-nitrobenzene, 98%, Thermo Scientific™
$71.90 - $205.73
Chemical Identifiers
CAS | 7087-65-2 |
---|---|
Molecular Formula | C6H3BrFNO2 |
Molecular Weight (g/mol) | 219.997 |
MDL Number | MFCD09955454 |
InChI Key | SWXVEPMSQBEVRH-UHFFFAOYSA-N |
Synonym | 3-bromo-5-fluoronitrobenzene, 1-nitro-3-fluoro-5-bromobenzene, benzene, 1-bromo-3-fluoro-5-nitro, pubchem17569, acmc-209ogb, 3-fluoro-5-nitrobromobenzene, 5-bromo-3-fluoro-1-nitrobenzene |
PubChem CID | 15020157 |
IUPAC Name | 1-bromo-3-fluoro-5-nitrobenzene |
SMILES | C1=C(C=C(C=C1F)Br)[N+](=O)[O-] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH61192MD
|
Thermo Scientific Chemicals
H61192MD |
250 mg |
N/A
|
|
|||||
AAH6119203
|
Thermo Scientific Chemicals
H6119203 |
1 g |
N/A
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
7087-65-2 | |
219.997 | |
SWXVEPMSQBEVRH-UHFFFAOYSA-N | |
15020157 | |
C1=C(C=C(C=C1F)Br)[N+](=O)[O-] |
C6H3BrFNO2 | |
MFCD09955454 | |
3-bromo-5-fluoronitrobenzene, 1-nitro-3-fluoro-5-bromobenzene, benzene, 1-bromo-3-fluoro-5-nitro, pubchem17569, acmc-209ogb, 3-fluoro-5-nitrobromobenzene, 5-bromo-3-fluoro-1-nitrobenzene | |
1-bromo-3-fluoro-5-nitrobenzene |
Specifications
7087-65-2 | |
C6H3BrFNO2 | |
250 mg | |
3-bromo-5-fluoronitrobenzene, 1-nitro-3-fluoro-5-bromobenzene, benzene, 1-bromo-3-fluoro-5-nitro, pubchem17569, acmc-209ogb, 3-fluoro-5-nitrobromobenzene, 5-bromo-3-fluoro-1-nitrobenzene | |
C1=C(C=C(C=C1F)Br)[N+](=O)[O-] | |
219.997 | |
220 | |
Liquid |
Yellow | |
MFCD09955454 | |
UN2810 | |
SWXVEPMSQBEVRH-UHFFFAOYSA-N | |
1-bromo-3-fluoro-5-nitrobenzene | |
15020157 | |
98% | |
1-Bromo-3-fluoro-5-nitrobenzene |
Safety and Handling
GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures