missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromo-3-iodobenzene, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1959614

 View more versions of this product

Catalog No. AAA1959614


Explore available promotions

Description

Description

1-Bromo-3-iodobenzene is used in the preparation of 3'-bromo-2,3,4,5,6-pentaethyl-biphenyl by reacting with hex-3-yne. It is also used to prepare 1-(3'-bromophenyl)-3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodec-1-ene and 1-(3'-bromophenyl)-3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooct-1-ene. Further, it is involved in the preparation of oxygen-tethered 1,6-enynes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-3-iodobenzene is used in the preparation of 3′-bromo-2,3,4,5,6-pentaethyl-biphenyl by reacting with hex-3-yne. It is also used to prepare 1-(3′-bromophenyl)-3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodec-1-ene and 1-(3′-bromophenyl)-3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooct-1-ene. Further, it is involved in the preparation of oxygen-tethered 1,6-enynes.

Solubility
Immiscible with water.

Notes
Light sensitive. Incompatible with strong oxidizing agents, strong bases and strong acids. Store in a cool place.
Specifications

Specifications

1-Bromo-3-iodobenzene
-9°C
252°C
Recognizable
1.662
25 g
Light sensitive
Immiscible with water.
BrC1=CC=CC(I)=C1
282.91
282.91
591-18-4
2.219
>110°C (230°F)
C6H4BrI
MFCD00001043
1854384
3-bromoiodobenzene, benzene, 1-bromo-3-iodo, m-bromoiodobenzene, 1-bromo-3-iodo-benzene, 3-bromo-1-iodobenzene, attercop-chm at127411, m-bromjodbenzol, 3-iodobromobenzene, 3-bromo iodobenzene
CTPUUDQIXKUAMO-UHFFFAOYSA-N
1-bromo-3-iodobenzene
11561
98%

Chemical Identifiers

591-18-4
282.91
CTPUUDQIXKUAMO-UHFFFAOYSA-N
11561
BrC1=CC=CC(I)=C1
C6H4BrI
MFCD00001043
3-bromoiodobenzene, benzene, 1-bromo-3-iodo, m-bromoiodobenzene, 1-bromo-3-iodo-benzene, 3-bromo-1-iodobenzene, attercop-chm at127411, m-bromjodbenzol, 3-iodobromobenzene, 3-bromo iodobenzene
1-bromo-3-iodobenzene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 209-703-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only