missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromo-3-iodobenzene, 98%, Thermo Scientific Chemicals

$159.62 - $611.77

Chemical Identifiers

CAS 591-18-4
Molecular Formula C6H4BrI
Molecular Weight (g/mol) 282.91
MDL Number MFCD00001043
InChI Key CTPUUDQIXKUAMO-UHFFFAOYSA-N
Synonym 3-bromoiodobenzene, benzene, 1-bromo-3-iodo, m-bromoiodobenzene, 1-bromo-3-iodo-benzene, 3-bromo-1-iodobenzene, attercop-chm at127411, m-bromjodbenzol, 3-iodobromobenzene, 3-bromo iodobenzene
PubChem CID 11561
IUPAC Name 1-bromo-3-iodobenzene
SMILES BrC1=CC=CC(I)=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1959614
View Documents
Thermo Scientific Chemicals
A1959614
25 g
Each for $159.62
 
AAA1959622
View Documents
Thermo Scientific Chemicals
A1959622
100 g
Each for $611.77
 
Description

Description

1-Bromo-3-iodobenzene is used in the preparation of 3'-bromo-2,3,4,5,6-pentaethyl-biphenyl by reacting with hex-3-yne. It is also used to prepare 1-(3'-bromophenyl)-3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodec-1-ene and 1-(3'-bromophenyl)-3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooct-1-ene. Further, it is involved in the preparation of oxygen-tethered 1,6-enynes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-3-iodobenzene is used in the preparation of 3′-bromo-2,3,4,5,6-pentaethyl-biphenyl by reacting with hex-3-yne. It is also used to prepare 1-(3′-bromophenyl)-3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodec-1-ene and 1-(3′-bromophenyl)-3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooct-1-ene. Further, it is involved in the preparation of oxygen-tethered 1,6-enynes.

Solubility
Immiscible with water.

Notes
Light sensitive. Incompatible with strong oxidizing agents, strong bases and strong acids. Store in a cool place.
Specifications

Chemical Identifiers

591-18-4
282.91
CTPUUDQIXKUAMO-UHFFFAOYSA-N
11561
BrC1=CC=CC(I)=C1
C6H4BrI
MFCD00001043
3-bromoiodobenzene, benzene, 1-bromo-3-iodo, m-bromoiodobenzene, 1-bromo-3-iodo-benzene, 3-bromo-1-iodobenzene, attercop-chm at127411, m-bromjodbenzol, 3-iodobromobenzene, 3-bromo iodobenzene
1-bromo-3-iodobenzene

Specifications

591-18-4
2.219
>110°C (230°F)
C6H4BrI
MFCD00001043
1854384
3-bromoiodobenzene, benzene, 1-bromo-3-iodo, m-bromoiodobenzene, 1-bromo-3-iodo-benzene, 3-bromo-1-iodobenzene, attercop-chm at127411, m-bromjodbenzol, 3-iodobromobenzene, 3-bromo iodobenzene
CTPUUDQIXKUAMO-UHFFFAOYSA-N
1-bromo-3-iodobenzene
11561
98%
-9°C
252°C
Recognizable
1.662
25 g
Light sensitive
Immiscible with water.
BrC1=CC=CC(I)=C1
282.91
282.91
1-Bromo-3-iodobenzene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 209-703-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only