missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromo-4-chlorobenzene, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1216022

 View more versions of this product

Catalog No. AAA1216022


Explore available promotions

Description

Description

1-Bromo-4-chlorobenzene is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-4-chlorobenzene is used as a pharmaceutical intermediate.

Solubility
Soluble in water 0.045g/L.

Notes
Store in cool, dry place, in well sealed container. Store away from oxidizing agents.
Specifications

Specifications

1-Bromo-4-chlorobenzene
106-39-8
196°C
Aromatic
MFCD00000600
1099014
Soluble in water 0.045g/L.
ClC1=CC=C(Br)C=C1
191.45
191.46
63°C to 67°C
1.651
100 g
C6H4BrCl
UN2811
4-bromochlorobenzene, p-bromochlorobenzene, 4-chlorobromobenzene, p-chlorobromobenzene, p-chlorophenyl bromide, benzene, 1-bromo-4-chloro, 4-chlorophenyl bromide, 1-chloro-4-bromobenzene, 4-chloro-1-bromobenzene, p-bromoclorobenzene
NHDODQWIKUYWMW-UHFFFAOYSA-N
1-bromo-4-chlorobenzene
7806
≥98%

Chemical Identifiers

106-39-8
191.45
NHDODQWIKUYWMW-UHFFFAOYSA-N
7806
ClC1=CC=C(Br)C=C1
C6H4BrCl
MFCD00000600
4-bromochlorobenzene, p-bromochlorobenzene, 4-chlorobromobenzene, p-chlorobromobenzene, p-chlorophenyl bromide, benzene, 1-bromo-4-chloro, 4-chlorophenyl bromide, 1-chloro-4-bromobenzene, 4-chloro-1-bromobenzene, p-bromoclorobenzene
1-bromo-4-chlorobenzene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 203-392-3

RTECSNumber : CY9006000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only