missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromo-4-methylcyclohexane, cis + trans, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L0264014

 View more versions of this product

Catalog No. AAL0264014



Description

Description

1-Bromo-4-methylcyclohexane is used as intermediates in pharmaceutical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-4-methylcyclohexane is used as intermediates in pharmaceutical research.

Solubility
Not miscible in water.

Notes
Store away from oxidizing agents. Incompatible with oxidizing agents.
Specifications

Specifications

1-Bromo-4-methylcyclohexane
6294-40-2
94°C to 96°C (50 mmHg)
1.488
25 g
cyclohexane, 1-bromo-4-methyl, p-methylcyclohexyl bromide, 4-bromo-1-methylcyclohexane, 4-methylcyclohexyl bromide, acmc-1b9h5, 1-bromo-4-methyl-cyclohexane, 4-methylcyclohexyl bromide cis+trans, cyclohexane, 1-bromo-4-methyl-, trans, 1-bromo-4-methylcyclohexane, cis + trans
RLOHLPHAOHGRNM-UHFFFAOYSA-N
1-bromo-4-methylcyclohexane
22727
97%
Cis + Trans
1.262
C7H13Br
MFCD00014282
1900954
Not miscible in water.
CC1CCC(CC1)Br
177.085
177.09

Chemical Identifiers

6294-40-2
177.085
RLOHLPHAOHGRNM-UHFFFAOYSA-N
22727
CC1CCC(CC1)Br
C7H13Br
MFCD00014282
cyclohexane, 1-bromo-4-methyl, p-methylcyclohexyl bromide, 4-bromo-1-methylcyclohexane, 4-methylcyclohexyl bromide, acmc-1b9h5, 1-bromo-4-methyl-cyclohexane, 4-methylcyclohexyl bromide cis+trans, cyclohexane, 1-bromo-4-methyl-, trans, 1-bromo-4-methylcyclohexane, cis + trans
1-bromo-4-methylcyclohexane
Safety and Handling

Safety and Handling

EINECSNumber : 228-559-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only