missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Bromo-4-methylcyclohexane, cis + trans, 97%, Thermo Scientific Chemicals
$141.34 - $554.85
Chemical Identifiers
CAS | 6294-40-2 |
---|---|
Molecular Formula | C7H13Br |
Molecular Weight (g/mol) | 177.085 |
MDL Number | MFCD00014282 |
InChI Key | RLOHLPHAOHGRNM-UHFFFAOYSA-N |
Synonym | cyclohexane, 1-bromo-4-methyl, p-methylcyclohexyl bromide, 4-bromo-1-methylcyclohexane, 4-methylcyclohexyl bromide, acmc-1b9h5, 1-bromo-4-methyl-cyclohexane, 4-methylcyclohexyl bromide cis+trans, cyclohexane, 1-bromo-4-methyl-, trans, 1-bromo-4-methylcyclohexane, cis + trans |
PubChem CID | 22727 |
IUPAC Name | 1-bromo-4-methylcyclohexane |
SMILES | CC1CCC(CC1)Br |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0264006
|
Thermo Scientific Chemicals
L0264006 |
5 g |
Each for $141.34
|
|
|||||
AAL0264014
|
Thermo Scientific Chemicals
L0264014 |
25 g |
Each for $554.85
|
|
|||||
Description
1-Bromo-4-methylcyclohexane is used as intermediates in pharmaceutical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Bromo-4-methylcyclohexane is used as intermediates in pharmaceutical research.
Solubility
Not miscible in water.
Notes
Store away from oxidizing agents. Incompatible with oxidizing agents.
Chemical Identifiers
6294-40-2 | |
177.085 | |
RLOHLPHAOHGRNM-UHFFFAOYSA-N | |
22727 | |
CC1CCC(CC1)Br |
C7H13Br | |
MFCD00014282 | |
cyclohexane, 1-bromo-4-methyl, p-methylcyclohexyl bromide, 4-bromo-1-methylcyclohexane, 4-methylcyclohexyl bromide, acmc-1b9h5, 1-bromo-4-methyl-cyclohexane, 4-methylcyclohexyl bromide cis+trans, cyclohexane, 1-bromo-4-methyl-, trans, 1-bromo-4-methylcyclohexane, cis + trans | |
1-bromo-4-methylcyclohexane |
Specifications
6294-40-2 | |
94°C to 96°C (50 mmHg) | |
1.488 | |
5 g | |
cyclohexane, 1-bromo-4-methyl, p-methylcyclohexyl bromide, 4-bromo-1-methylcyclohexane, 4-methylcyclohexyl bromide, acmc-1b9h5, 1-bromo-4-methyl-cyclohexane, 4-methylcyclohexyl bromide cis+trans, cyclohexane, 1-bromo-4-methyl-, trans, 1-bromo-4-methylcyclohexane, cis + trans | |
RLOHLPHAOHGRNM-UHFFFAOYSA-N | |
1-bromo-4-methylcyclohexane | |
22727 | |
97% |
1.262 | |
C7H13Br | |
MFCD00014282 | |
1900954 | |
Not miscible in water. | |
CC1CCC(CC1)Br | |
177.085 | |
177.09 | |
1-Bromo-4-methylcyclohexane, Cis + Trans |
Safety and Handling
EINECSNumber : 228-559-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only