missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromo-6-chlorohexane, 97%, Thermo Scientific Chemicals

$63.55 - $180.42

Chemical Identifiers

CAS 6294-17-3
Molecular Formula C6H12BrCl
Molecular Weight (g/mol) 199.52
MDL Number MFCD00001019
InChI Key JTYUIAOHIYZBPB-UHFFFAOYSA-N
Synonym 6-chlorohexyl bromide, hexane, 1-bromo-6-chloro, 1-chloro-6-bromohexane, hexyl bromide, 6-chloro, 6-bromohexyl chloride, 1-bromo-6-chloro hexane, 1-bromo-6-chloro-hexane, acmc-1b9m0, 1-bromo-6-chlorohexane, jtyuiaohiyzbpb-uhfffaoysa
PubChem CID 80516
IUPAC Name 1-bromo-6-chlorohexane
SMILES ClCCCCCCBr
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2161906
View Documents
Thermo Scientific Chemicals
B2161906
5 g N/A
 
AAB2161914
View Documents
Thermo Scientific Chemicals
B2161914
25 g N/A
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6294-17-3
199.52
JTYUIAOHIYZBPB-UHFFFAOYSA-N
80516
ClCCCCCCBr
C6H12BrCl
MFCD00001019
6-chlorohexyl bromide, hexane, 1-bromo-6-chloro, 1-chloro-6-bromohexane, hexyl bromide, 6-chloro, 6-bromohexyl chloride, 1-bromo-6-chloro hexane, 1-bromo-6-chloro-hexane, acmc-1b9m0, 1-bromo-6-chlorohexane, jtyuiaohiyzbpb-uhfffaoysa
1-bromo-6-chlorohexane

Specifications

6294-17-3
109°C to 110°C (2 mmHg)
C6H12BrCl
MFCD00001019
6-chlorohexyl bromide, hexane, 1-bromo-6-chloro, 1-chloro-6-bromohexane, hexyl bromide, 6-chloro, 6-bromohexyl chloride, 1-bromo-6-chloro hexane, 1-bromo-6-chloro-hexane, acmc-1b9m0, 1-bromo-6-chlorohexane, jtyuiaohiyzbpb-uhfffaoysa
ClCCCCCCBr
199.52
199.53
1-Bromo-6-chlorohexane
1.337
101°C (214°F)
1.4806
5 g
JTYUIAOHIYZBPB-UHFFFAOYSA-N
1-bromo-6-chlorohexane
80516
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 228-555-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only