missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(Bromomethyl)naphthalene, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6411614

 View more versions of this product

Catalog No. AAH6411614


Explore available promotions

Description

Description

It is employed as a intermediate for organic synthesis and pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediate for organic synthesis and pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Specifications

Specifications

1-(Bromomethyl)naphthalene
White
213°C (100 mmHg)
1.66
25 g
742417
Slightly soluble in water.
C1=CC=C2C(=C1)C=CC=C2CBr
221.097
221.1
Powder
3163-27-7
1.44 g/mL
C11H9Br
MFCD00010804
UN3261
1-bromomethyl naphthalene, 1-bromomethyl-naphthalene, naphthalene, 1-bromomethyl, 1-bromomethy naphthalene, naphtylmethylbromide, bromomethylnaphthalene, naphthylmethyl bromide, pubchem9389
RZJGKPNCYQZFGR-UHFFFAOYSA-N
1-(bromomethyl)naphthalene
137844
98%

Chemical Identifiers

3163-27-7
221.097
RZJGKPNCYQZFGR-UHFFFAOYSA-N
137844
C1=CC=C2C(=C1)C=CC=C2CBr
C11H9Br
MFCD00010804
1-bromomethyl naphthalene, 1-bromomethyl-naphthalene, naphthalene, 1-bromomethyl, 1-bromomethy naphthalene, naphtylmethylbromide, bromomethylnaphthalene, naphthylmethyl bromide, pubchem9389
1-(bromomethyl)naphthalene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 801-817-2

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only