Learn More
1-(Bromomethyl)naphthalene, 98%, Thermo Scientific Chemicals
$245.53 - $245.53
Chemical Identifiers
CAS | 3163-27-7 |
---|---|
Molecular Formula | C11H9Br |
Molecular Weight (g/mol) | 221.097 |
MDL Number | MFCD00010804 |
InChI Key | RZJGKPNCYQZFGR-UHFFFAOYSA-N |
Synonym | 1-bromomethyl naphthalene, 1-bromomethyl-naphthalene, naphthalene, 1-bromomethyl, 1-bromomethy naphthalene, naphtylmethylbromide, bromomethylnaphthalene, naphthylmethyl bromide, pubchem9389 |
PubChem CID | 137844 |
IUPAC Name | 1-(bromomethyl)naphthalene |
SMILES | C1=CC=C2C(=C1)C=CC=C2CBr |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6411614
|
Thermo Scientific Chemicals
H6411614 |
25 g |
N/A
|
|
|||||
Description
It is employed as a intermediate for organic synthesis and pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is employed as a intermediate for organic synthesis and pharmaceutical.
Solubility
Slightly soluble in water.
Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Chemical Identifiers
3163-27-7 | |
221.097 | |
RZJGKPNCYQZFGR-UHFFFAOYSA-N | |
137844 | |
C1=CC=C2C(=C1)C=CC=C2CBr |
C11H9Br | |
MFCD00010804 | |
1-bromomethyl naphthalene, 1-bromomethyl-naphthalene, naphthalene, 1-bromomethyl, 1-bromomethy naphthalene, naphtylmethylbromide, bromomethylnaphthalene, naphthylmethyl bromide, pubchem9389 | |
1-(bromomethyl)naphthalene |
Specifications
3163-27-7 | |
1.44 g/mL | |
C11H9Br | |
MFCD00010804 | |
UN3261 | |
1-bromomethyl naphthalene, 1-bromomethyl-naphthalene, naphthalene, 1-bromomethyl, 1-bromomethy naphthalene, naphtylmethylbromide, bromomethylnaphthalene, naphthylmethyl bromide, pubchem9389 | |
RZJGKPNCYQZFGR-UHFFFAOYSA-N | |
1-(bromomethyl)naphthalene | |
137844 | |
98% | |
1-(Bromomethyl)naphthalene |
White | |
213°C (100 mmHg) | |
1.66 | |
25 g | |
742417 | |
Slightly soluble in water. | |
C1=CC=C2C(=C1)C=CC=C2CBr | |
221.097 | |
221.1 | |
Powder |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 801-817-2
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only