missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromooctane, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A147220I

 View more versions of this product

Catalog No. AAA147220I


Explore available promotions

Description

Description

1-Bromooctane is used in organic synthesis. It is also used as an extraction solvent in the determination of volatile organic compounds present in water samples. Further, it is used in the preparation of quaternary ammonium chloride-modified poly(propylenimine) dendrimers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromooctane is used in organic synthesis. It is also used as an extraction solvent in the determination of volatile organic compounds present in water samples. Further, it is used in the preparation of quaternary ammonium chloride-modified poly(propylenimine) dendrimers.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents and bases.
Specifications

Specifications

1-Bromooctane
-55°C
200°C to 202°C
Characteristic
1.4518
MFCD00000276
UN3082
14,6764
Immiscible with water.
CCCCCCCCBr
193.13
193.13
111-83-1
1.114
78°C (172°F)
C8H17Br
CH3(CH2)7Br
5000 g
1733136
n-octyl bromide, octyl bromide, 1-octylbromide, octane, 1-bromo, bromooctane, 1-bromo-octane, 1-bromo octane, 1-bromo-n-octane, unii-q382hpc6lc, octane, bromo
VMKOFRJSULQZRM-UHFFFAOYSA-N
1-bromooctane
8140
≥98%

Chemical Identifiers

111-83-1
193.13
VMKOFRJSULQZRM-UHFFFAOYSA-N
8140
CCCCCCCCBr
C8H17Br
MFCD00000276
n-octyl bromide, octyl bromide, 1-octylbromide, octane, 1-bromo, bromooctane, 1-bromo-octane, 1-bromo octane, 1-bromo-n-octane, unii-q382hpc6lc, octane, bromo
1-bromooctane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

P210-P280-P370+P378q-P501c

H227

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.

EINECSNumber : 203-912-9

RTECSNumber : RG8575000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only