Learn More
1-Bromooctane, 98+%, Thermo Scientific Chemicals
$75.25 - $754.21
Chemical Identifiers
CAS | 111-83-1 |
---|---|
Molecular Formula | C8H17Br |
Molecular Weight (g/mol) | 193.13 |
MDL Number | MFCD00000276 |
InChI Key | VMKOFRJSULQZRM-UHFFFAOYSA-N |
Synonym | n-octyl bromide, octyl bromide, 1-octylbromide, octane, 1-bromo, bromooctane, 1-bromo-octane, 1-bromo octane, 1-bromo-n-octane, unii-q382hpc6lc, octane, bromo |
PubChem CID | 8140 |
IUPAC Name | 1-bromooctane |
SMILES | CCCCCCCCBr |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA1472230
|
Thermo Scientific Chemicals
A1472230 |
250 g |
N/A
|
|
||||||
AAA147220B
|
Thermo Scientific Chemicals
A147220B |
1000 g |
N/A
|
|
||||||
AAA147220I
|
Thermo Scientific Chemicals
A147220I |
5000 g |
N/A
|
|
||||||
Description
1-Bromooctane is used in organic synthesis. It is also used as an extraction solvent in the determination of volatile organic compounds present in water samples. Further, it is used in the preparation of quaternary ammonium chloride-modified poly(propylenimine) dendrimers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Bromooctane is used in organic synthesis. It is also used as an extraction solvent in the determination of volatile organic compounds present in water samples. Further, it is used in the preparation of quaternary ammonium chloride-modified poly(propylenimine) dendrimers.
Solubility
Immiscible with water.
Notes
Incompatible with strong oxidizing agents and bases.
Chemical Identifiers
111-83-1 | |
193.13 | |
VMKOFRJSULQZRM-UHFFFAOYSA-N | |
8140 | |
CCCCCCCCBr |
C8H17Br | |
MFCD00000276 | |
n-octyl bromide, octyl bromide, 1-octylbromide, octane, 1-bromo, bromooctane, 1-bromo-octane, 1-bromo octane, 1-bromo-n-octane, unii-q382hpc6lc, octane, bromo | |
1-bromooctane |
Specifications
111-83-1 | |
1.114 | |
78°C (172°F) | |
C8H17Br | |
CH3(CH2)7Br | |
250 g | |
1733136 | |
n-octyl bromide, octyl bromide, 1-octylbromide, octane, 1-bromo, bromooctane, 1-bromo-octane, 1-bromo octane, 1-bromo-n-octane, unii-q382hpc6lc, octane, bromo | |
VMKOFRJSULQZRM-UHFFFAOYSA-N | |
1-bromooctane | |
8140 | |
≥98% |
-55°C | |
200°C to 202°C | |
Characteristic | |
1.4518 | |
MFCD00000276 | |
UN3082 | |
14,6764 | |
Immiscible with water. | |
CCCCCCCCBr | |
193.13 | |
193.13 | |
1-Bromooctane |
Safety and Handling
GHS H Statement
H227
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber : 203-912-9
RTECSNumber : RG8575000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only