missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Bromoperfluorononane, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals L1423106

 View more versions of this product

Catalog No. AAL1423106

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
1-Bromoperfluorononane is a specialty product for proteomics research applications. It is also used as pharmaceutical intermediate and in chemical research.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, alkali metals and alkaline earth metals.
Specifications

Specifications

1-Bromoperfluorononane
27°C to 30°C
159°C
C9BrF19
MFCD00153257
1-bromoperfluorononane, 1-bromononadecafluorononane, perfluorononyl bromide, nonadecafluorononyl bromide, acmc-1azu3, 1-bromononadecafluorononane1-bromoperfluorononane, 1-bromononadecafluorononane; 1-bromoperfluorononane, 1-bromanyl-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-nonadecakis fluoranyl nonane
OWIAIPIQXHPUHV-UHFFFAOYSA-N
1-bromo-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-nonadecafluorononane
2776260
98%
558-96-3
1.9
None
CF3(CF2)8Br
5 g
Insoluble in water.
C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(C(C(C(C(F)(F)Br)(F)F)(F)F)(F)F)(F)F
548.973
548.97

Chemical Identifiers

558-96-3
548.973
OWIAIPIQXHPUHV-UHFFFAOYSA-N
2776260
C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(C(C(C(C(F)(F)Br)(F)F)(F)F)(F)F)(F)F
C9BrF19
MFCD00153257
1-bromoperfluorononane, 1-bromononadecafluorononane, perfluorononyl bromide, nonadecafluorononyl bromide, acmc-1azu3, 1-bromononadecafluorononane1-bromoperfluorononane, 1-bromononadecafluorononane; 1-bromoperfluorononane, 1-bromanyl-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-nonadecakis fluoranyl nonane
1-bromo-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-nonadecafluorononane
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions