missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Bromoperfluorononane, 98%, Thermo Scientific™
$43.68 - $141.34
Chemical Identifiers
CAS | 558-96-3 |
---|---|
Molecular Formula | C9BrF19 |
Molecular Weight (g/mol) | 548.973 |
MDL Number | MFCD00153257 |
InChI Key | OWIAIPIQXHPUHV-UHFFFAOYSA-N |
Synonym | 1-bromoperfluorononane, 1-bromononadecafluorononane, perfluorononyl bromide, nonadecafluorononyl bromide, acmc-1azu3, 1-bromononadecafluorononane1-bromoperfluorononane, 1-bromononadecafluorononane; 1-bromoperfluorononane, 1-bromanyl-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-nonadecakis fluoranyl nonane |
PubChem CID | 2776260 |
IUPAC Name | 1-bromo-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-nonadecafluorononane |
SMILES | C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(C(C(C(C(F)(F)Br)(F)F)(F)F)(F)F)(F)F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1423103
|
Thermo Scientific Chemicals
L1423103 |
1 g |
Each for $43.68
|
|
|||||
AAL1423106
|
Thermo Scientific Chemicals
L1423106 |
5 g |
Each for $141.34
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications1-Bromoperfluorononane is a specialty product for proteomics research applications. It is also used as pharmaceutical intermediate and in chemical research.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, alkali metals and alkaline earth metals.
Chemical Identifiers
558-96-3 | |
548.973 | |
OWIAIPIQXHPUHV-UHFFFAOYSA-N | |
2776260 | |
C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(C(C(C(C(F)(F)Br)(F)F)(F)F)(F)F)(F)F |
C9BrF19 | |
MFCD00153257 | |
1-bromoperfluorononane, 1-bromononadecafluorononane, perfluorononyl bromide, nonadecafluorononyl bromide, acmc-1azu3, 1-bromononadecafluorononane1-bromoperfluorononane, 1-bromononadecafluorononane; 1-bromoperfluorononane, 1-bromanyl-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-nonadecakis fluoranyl nonane | |
1-bromo-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-nonadecafluorononane |
Specifications
558-96-3 | |
1.9 | |
None | |
CF3(CF2)8Br | |
1 g | |
Insoluble in water. | |
C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(C(C(C(C(F)(F)Br)(F)F)(F)F)(F)F)(F)F | |
548.973 | |
548.97 | |
1-Bromoperfluorononane |
27°C to 30°C | |
159°C | |
C9BrF19 | |
MFCD00153257 | |
1-bromoperfluorononane, 1-bromononadecafluorononane, perfluorononyl bromide, nonadecafluorononyl bromide, acmc-1azu3, 1-bromononadecafluorononane1-bromoperfluorononane, 1-bromononadecafluorononane; 1-bromoperfluorononane, 1-bromanyl-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-nonadecakis fluoranyl nonane | |
OWIAIPIQXHPUHV-UHFFFAOYSA-N | |
1-bromo-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-nonadecafluorononane | |
2776260 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures