missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Butanesulfonyl chloride, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 403320250

 View more versions of this product

Catalog No. AC403320250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Butanesulfonyl chloride
2386-60-9
100.0
1.2000g/mL
79°C
97.5% min. (GC)
C4H9ClO2S
CH3(CH2)3SO2Cl
25 g
1-butanesulfonyl chloride, butylsulfonyl chloride, butanesulfonyl chloride, 1-butanesulfonylchloride, 1-butylsulfonyl chloride, n-butanesulphonyl chloride, butanesulfonylchloride, butane-1-sulphonyl chloride, n-butylsulfonyl chloride, n-butanesulfonyl chloride
WEDIIKBPDQQQJU-UHFFFAOYSA-N
butane-1-sulfonyl chloride
75430
98%
99%
97.5
Colorless to Yellow or Orange
96.0°C (21.0 mmHg)
Authentic
Glass bottle
1.4540 to 1.4565
MFCD00007464
1.2
Solubility in water: decomposes.
CCCCS(=O)(=O)Cl
156.63
156.63
Liquid

Chemical Identifiers

2386-60-9
156.63
WEDIIKBPDQQQJU-UHFFFAOYSA-N
75430
CCCCS(=O)(=O)Cl
C4H9ClO2S
MFCD00007464
1-butanesulfonyl chloride, butylsulfonyl chloride, butanesulfonyl chloride, 1-butanesulfonylchloride, 1-butylsulfonyl chloride, n-butanesulphonyl chloride, butanesulfonylchloride, butane-1-sulphonyl chloride, n-butylsulfonyl chloride, n-butanesulfonyl chloride
butane-1-sulfonyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 219-204-8

RTECSNumber : EK5950000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only