missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Butanesulfonyl chloride, 98%, Thermo Scientific Chemicals

$150.70 - $407.10

Chemical Identifiers

CAS 2386-60-9
Molecular Formula C4H9ClO2S
Molecular Weight (g/mol) 156.63
MDL Number MFCD00007464
InChI Key WEDIIKBPDQQQJU-UHFFFAOYSA-N
Synonym 1-butanesulfonyl chloride, butylsulfonyl chloride, butanesulfonyl chloride, 1-butanesulfonylchloride, 1-butylsulfonyl chloride, n-butanesulphonyl chloride, butanesulfonylchloride, butane-1-sulphonyl chloride, n-butylsulfonyl chloride, n-butanesulfonyl chloride
PubChem CID 75430
IUPAC Name butane-1-sulfonyl chloride
SMILES CCCCS(=O)(=O)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC403320250
SDS View Documents
Thermo Scientific Chemicals
403320250
25 g Glass bottle
Each for $150.70
 
AC403321000
SDS View Documents
Thermo Scientific Chemicals
403321000
100 g Glass bottle
Each for $407.10
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2386-60-9
156.63
WEDIIKBPDQQQJU-UHFFFAOYSA-N
75430
CCCCS(=O)(=O)Cl
C4H9ClO2S
MFCD00007464
1-butanesulfonyl chloride, butylsulfonyl chloride, butanesulfonyl chloride, 1-butanesulfonylchloride, 1-butylsulfonyl chloride, n-butanesulphonyl chloride, butanesulfonylchloride, butane-1-sulphonyl chloride, n-butylsulfonyl chloride, n-butanesulfonyl chloride
butane-1-sulfonyl chloride

Specifications

2386-60-9
100.0
1.2000g/mL
79°C
97.5% min. (GC)
C4H9ClO2S
CH3(CH2)3SO2Cl
25 g
1-butanesulfonyl chloride, butylsulfonyl chloride, butanesulfonyl chloride, 1-butanesulfonylchloride, 1-butylsulfonyl chloride, n-butanesulphonyl chloride, butanesulfonylchloride, butane-1-sulphonyl chloride, n-butylsulfonyl chloride, n-butanesulfonyl chloride
WEDIIKBPDQQQJU-UHFFFAOYSA-N
butane-1-sulfonyl chloride
75430
98%
1-Butanesulfonyl chloride, 99%
97.5
Colorless to Yellow or Orange
96.0°C (21.0 mmHg)
Authentic
Glass bottle
1.4540 to 1.4565
MFCD00007464
1.2
Solubility in water: decomposes.
CCCCS(=O)(=O)Cl
156.63
156.63
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 219-204-8

RTECSNumber : EK5950000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only