missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Butanethiol, 99+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 220622500

 View more versions of this product

Catalog No. AC220622500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Butanethiol
109-79-5
100.0
Colorless
98.0°C
Authentic
Glass bottle
1.4420 to 1.4440
MFCD00004905
01, 370
0.84
1-butanethiol, butanethiol, butyl mercaptan, n-butyl mercaptan, n-butanethiol, butylthiol, thiobutyl alcohol, n-butylmercaptan, 1-mercaptobutane, 1-butyl mercaptan
WQAQPCDUOCURKW-UHFFFAOYSA-N
butane-1-thiol
8012
99+%
99+%
99.0
-116.0°C
0.8400g/mL
0.5°C
99+%
C4H10S
CH3(CH2)3SH
250 mL
02,53; 04,64
15, 1580
Solubility in water: slightly soluble. Other solubilities: soluble in alcohol and ether
CCCCS
90.18
90.18
Liquid

Chemical Identifiers

109-79-5
90.18
WQAQPCDUOCURKW-UHFFFAOYSA-N
8012
CCCCS
C4H10S
MFCD00004905
1-butanethiol, butanethiol, butyl mercaptan, n-butyl mercaptan, n-butanethiol, butylthiol, thiobutyl alcohol, n-butylmercaptan, 1-mercaptobutane, 1-butyl mercaptan
butane-1-thiol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Highly flammable liquid and vapor.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Danger

EINECSNumber : 203-705-3

RTECSNumber : EK6300000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only