missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Butanethiol, 99+%, Thermo Scientific Chemicals

$43.63 - $240.58

Chemical Identifiers

CAS 109-79-5
Molecular Formula C4H10S
Molecular Weight (g/mol) 90.18
MDL Number MFCD00004905
InChI Key WQAQPCDUOCURKW-UHFFFAOYSA-N
Synonym 1-butanethiol, butanethiol, butyl mercaptan, n-butyl mercaptan, n-butanethiol, butylthiol, thiobutyl alcohol, n-butylmercaptan, 1-mercaptobutane, 1-butyl mercaptan
PubChem CID 8012
IUPAC Name butane-1-thiol
SMILES CCCCS
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC220620050
SDS View Documents
Thermo Scientific Chemicals
220620050
5 mL Glass bottle
Each for $44.46
 
AC220620500
SDS View Documents
Thermo Scientific Chemicals
220620500
50 mL Glass bottle
Each for $43.63
 
AC220622500
SDS View Documents
Thermo Scientific Chemicals
220622500
250 mL Glass bottle
Each for $75.90
 
AC220620010
SDS View Documents
Thermo Scientific Chemicals
220620010
1 L Glass bottle
Each for $240.58
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

109-79-5
90.18
WQAQPCDUOCURKW-UHFFFAOYSA-N
8012
CCCCS
C4H10S
MFCD00004905
1-butanethiol, butanethiol, butyl mercaptan, n-butyl mercaptan, n-butanethiol, butylthiol, thiobutyl alcohol, n-butylmercaptan, 1-mercaptobutane, 1-butyl mercaptan
butane-1-thiol

Specifications

109-79-5
100.0
Colorless
98.0°C
Authentic
Glass bottle
1.4420 to 1.4440
MFCD00004905
01, 370
0.84
1-butanethiol, butanethiol, butyl mercaptan, n-butyl mercaptan, n-butanethiol, butylthiol, thiobutyl alcohol, n-butylmercaptan, 1-mercaptobutane, 1-butyl mercaptan
WQAQPCDUOCURKW-UHFFFAOYSA-N
butane-1-thiol
8012
99+%
1-Butanethiol, 99+%
99.0
-116.0°C
0.8400g/mL
0.5°C
99+%
C4H10S
CH3(CH2)3SH
5 mL
02,53; 04,64
15, 1580
Solubility in water: slightly soluble. Other solubilities: soluble in alcohol and ether
CCCCS
90.18
90.18
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Highly flammable liquid and vapor.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Danger

EINECSNumber : 203-705-3

RTECSNumber : EK6300000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only