missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-(Carbamoylmethyl)cyclohexaneacetic Acid 98.0+%, TCI America™
$111.06 - $324.76
Chemical Identifiers
CAS | 99189-60-3 |
---|---|
Molecular Formula | C10H17NO3 |
Molecular Weight (g/mol) | 199.25 |
MDL Number | MFCD02181086 |
InChI Key | QJGSJXLCJRXTRY-UHFFFAOYSA-N |
Synonym | 1,1-cyclohexanediacetic acid monoamide, 1,1-cyclohexanediacetic acid mono amide, 2-1-2-amino-2-oxoethyl cyclohexyl acetic acid, 1-carbamoylmethyl cyclohexaneacetic acid, 3,3-pentamethyleneglutaramic acid, 1-2-amino-2-oxoethyl cyclohexyl acetic acid, 1-2-amino-2-oxoethyl cyclohexaneacetic acid, cyclohexaneacetic acid, 1-2-amino-2-oxoethyl, 1-carbamoylmethyl-cyclohexyl-acetic acid |
PubChem CID | 665664 |
IUPAC Name | 2-[1-(2-amino-2-oxoethyl)cyclohexyl]acetic acid |
SMILES | C1CCC(CC1)(CC(=O)N)CC(=O)O |
Chemical Identifiers
99189-60-3 | |
199.25 | |
QJGSJXLCJRXTRY-UHFFFAOYSA-N | |
665664 | |
C1CCC(CC1)(CC(=O)N)CC(=O)O |
C10H17NO3 | |
MFCD02181086 | |
1,1-cyclohexanediacetic acid monoamide, 1,1-cyclohexanediacetic acid mono amide, 2-1-2-amino-2-oxoethyl cyclohexyl acetic acid, 1-carbamoylmethyl cyclohexaneacetic acid, 3,3-pentamethyleneglutaramic acid, 1-2-amino-2-oxoethyl cyclohexyl acetic acid, 1-2-amino-2-oxoethyl cyclohexaneacetic acid, cyclohexaneacetic acid, 1-2-amino-2-oxoethyl, 1-carbamoylmethyl-cyclohexyl-acetic acid | |
2-[1-(2-amino-2-oxoethyl)cyclohexyl]acetic acid |
Specifications
99189-60-3 | |
White | |
MFCD02181086 | |
1,1-cyclohexanediacetic acid monoamide, 1,1-cyclohexanediacetic acid mono amide, 2-1-2-amino-2-oxoethyl cyclohexyl acetic acid, 1-carbamoylmethyl cyclohexaneacetic acid, 3,3-pentamethyleneglutaramic acid, 1-2-amino-2-oxoethyl cyclohexyl acetic acid, 1-2-amino-2-oxoethyl cyclohexaneacetic acid, cyclohexaneacetic acid, 1-2-amino-2-oxoethyl, 1-carbamoylmethyl-cyclohexyl-acetic acid | |
C1CCC(CC1)(CC(=O)N)CC(=O)O | |
199.25 | |
199.25 | |
Crystalline Powder |
147°C | |
C10H17NO3 | |
5 g | |
QJGSJXLCJRXTRY-UHFFFAOYSA-N | |
2-[1-(2-amino-2-oxoethyl)cyclohexyl]acetic acid | |
665664 | |
≥98.0% (T) | |
1-(Carbamoylmethyl)cyclohexaneacetic Acid |
Safety and Handling
TSCA : No