missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Chloro-2-iodoethane, 96%, stab. with copper, Thermo Scientific Chemicals

$80.89 - $980.66

Chemical Identifiers

CAS 624-70-4
Molecular Formula C2H4ClI
Molecular Weight (g/mol) 190.41
MDL Number MFCD00041453
InChI Key JTWWWQGSFTWWDL-UHFFFAOYSA-N
Synonym 1,2-chloroiodoethane, 1-chloro-2-iodo-ethane, ethane, 1-chloro-2-iodo, 2-chloroethyl iodide, 2-iodoethyl chloride, 1-iodo-2-chloroethane, 2-chlor-1-jod-athan, 2-chloro-1-iodoethane, 2-chloro-1-iodo-ethane, acmc-1b3nb
PubChem CID 136438
IUPAC Name 1-chloro-2-iodoethane
SMILES ClCCI
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1074203
View Documents
Thermo Scientific Chemicals
L1074203
1 g
Each for $80.89
 
AAL1074206
View Documents
Thermo Scientific Chemicals
L1074206
5 g
Each for $257.49
 
AAL1074214
View Documents
Thermo Scientific Chemicals
L1074214
25 g
Each for $980.66
 
Description

Description

1-Chloro-2-iodoethane is used as a mild electrophile for iodination of metallated species, as in the conversion of an ortho-lithiated benzyl ether to the corresponding ortho-iodide. It reacts with 5-hydroxy-[1,4]naphthoquinone, and obtain the 5-(2-chloro-ethoxy)-[1,4]naphthoquinone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-2-iodoethane is used as a mild electrophile for iodination of metallated species, as in the conversion of an ortho-lithiated benzyl ether to the corresponding ortho-iodide. It reacts with 5-hydroxy-[1,4]naphthoquinone, and obtain the 5-(2-chloro-ethoxy)-[1,4]naphthoquinone.

Solubility
Soluble inorganic solvents. Insoluble in water.

Notes
Incompatible with strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

624-70-4
190.41
JTWWWQGSFTWWDL-UHFFFAOYSA-N
136438
ClCCI
C2H4ClI
MFCD00041453
1,2-chloroiodoethane, 1-chloro-2-iodo-ethane, ethane, 1-chloro-2-iodo, 2-chloroethyl iodide, 2-iodoethyl chloride, 1-iodo-2-chloroethane, 2-chlor-1-jod-athan, 2-chloro-1-iodoethane, 2-chloro-1-iodo-ethane, acmc-1b3nb
1-chloro-2-iodoethane

Specifications

624-70-4
61°C to 62°C (30 mmHg)
C2H4ClI
MFCD00041453
1730865
1,2-chloroiodoethane, 1-chloro-2-iodo-ethane, ethane, 1-chloro-2-iodo, 2-chloroethyl iodide, 2-iodoethyl chloride, 1-iodo-2-chloroethane, 2-chlor-1-jod-athan, 2-chloro-1-iodoethane, 2-chloro-1-iodo-ethane, acmc-1b3nb
JTWWWQGSFTWWDL-UHFFFAOYSA-N
1-chloro-2-iodoethane
136438
96%
2.088
>100°C (212°F)
1.566
1 g
Light sensitive
Soluble inorganic solvents. Insoluble in water.
ClCCI
190.41
190.41
1-Chloro-2-iodoethane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 677-827-4

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only