missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Chloro-2-methyl-2-phenylpropane, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 124340050

 View more versions of this product

Catalog No. AC124340050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

1-Chloro-2-methyl-2-phenylpropane
515-40-2
100.0
95°C to 100°C (10.0mmHg)
Authentic
Glass bottle
1.5230 to 1.525
MFCD00000940
05,II,32
15,6544
Solubility in water: insoluble
CC(C)(CCl)C1=CC=CC=C1
168.66
168.67
98%
97.5
1.0400g/mL
96°C
97.5% min. (GC)
C10H13Cl
C6H5C(CH3)2CH2Cl
5mL
1.04
neophyl chloride, 1-chloro-2-methyl-2-phenylpropane, 1-chloro-2-methylpropan-2-yl benzene, benzene, 2-chloro-1,1-dimethylethyl, 2-methyl-2-phenylpropyl chloride, neophylchloride, 2-chloro-1,1-dimethylethyl benzene, unii-of6e0410nf, beta-chloro-tert-butylbenzene, 2-chloromethyl-2-phenylpropane
DNXXUUPUQXSUFH-UHFFFAOYSA-N
(1-chloro-2-methylpropan-2-yl)benzene
68191
98%

Chemical Identifiers

515-40-2
168.66
DNXXUUPUQXSUFH-UHFFFAOYSA-N
68191
CC(C)(CCl)C1=CC=CC=C1
C10H13Cl
MFCD00000940
neophyl chloride, 1-chloro-2-methyl-2-phenylpropane, 1-chloro-2-methylpropan-2-yl benzene, benzene, 2-chloro-1,1-dimethylethyl, 2-methyl-2-phenylpropyl chloride, neophylchloride, 2-chloro-1,1-dimethylethyl benzene, unii-of6e0410nf, beta-chloro-tert-butylbenzene, 2-chloromethyl-2-phenylpropane
(1-chloro-2-methylpropan-2-yl)benzene
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 208-197-7

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only