missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Chloro-2-methyl-2-phenylpropane, 98%, Thermo Scientific™

Chemical Identifiers

CAS 515-40-2
Molecular Formula C10H13Cl
Molecular Weight (g/mol) 168.66
MDL Number MFCD00000940
InChI Key DNXXUUPUQXSUFH-UHFFFAOYSA-N
Synonym neophyl chloride, 1-chloro-2-methyl-2-phenylpropane, 1-chloro-2-methylpropan-2-yl benzene, benzene, 2-chloro-1,1-dimethylethyl, 2-methyl-2-phenylpropyl chloride, neophylchloride, 2-chloro-1,1-dimethylethyl benzene, unii-of6e0410nf, beta-chloro-tert-butylbenzene, 2-chloromethyl-2-phenylpropane
PubChem CID 68191
IUPAC Name (1-chloro-2-methylpropan-2-yl)benzene
SMILES CC(C)(CCl)C1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC124340050
SDS View Documents
Thermo Scientific Chemicals
124340050
5mL Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

515-40-2
168.66
DNXXUUPUQXSUFH-UHFFFAOYSA-N
68191
CC(C)(CCl)C1=CC=CC=C1
C10H13Cl
MFCD00000940
neophyl chloride, 1-chloro-2-methyl-2-phenylpropane, 1-chloro-2-methylpropan-2-yl benzene, benzene, 2-chloro-1,1-dimethylethyl, 2-methyl-2-phenylpropyl chloride, neophylchloride, 2-chloro-1,1-dimethylethyl benzene, unii-of6e0410nf, beta-chloro-tert-butylbenzene, 2-chloromethyl-2-phenylpropane
(1-chloro-2-methylpropan-2-yl)benzene

Specifications

515-40-2
100.0
95°C to 100°C (10.0mmHg)
Authentic
Glass bottle
1.5230 to 1.525
MFCD00000940
05,II,32
15,6544
Solubility in water: insoluble
CC(C)(CCl)C1=CC=CC=C1
168.66
168.67
1-Chloro-2-methyl-2-phenylpropane, 98%
97.5
1.0400g/mL
96°C
97.5% min. (GC)
C10H13Cl
C6H5C(CH3)2CH2Cl
5mL
1.04
neophyl chloride, 1-chloro-2-methyl-2-phenylpropane, 1-chloro-2-methylpropan-2-yl benzene, benzene, 2-chloro-1,1-dimethylethyl, 2-methyl-2-phenylpropyl chloride, neophylchloride, 2-chloro-1,1-dimethylethyl benzene, unii-of6e0410nf, beta-chloro-tert-butylbenzene, 2-chloromethyl-2-phenylpropane
DNXXUUPUQXSUFH-UHFFFAOYSA-N
(1-chloro-2-methylpropan-2-yl)benzene
68191
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 208-197-7

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only