Learn More
1-Chloro-5-iodopentane, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L0446909
Description
1-Chloro-5-iodopentane is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Chloro-5-iodopentane is used as a pharmaceutical intermediate.
Solubility
Not miscible or difficult to mix with water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to light.
Specifications
1-Chloro-5-iodopentane | |
1.663 | |
>110°C (230°F) | |
1.5305 | |
10 g | |
Light sensitive | |
Not miscible or difficult to mix with water. | |
C(CCCl)CCI | |
232.489 | |
232.5 |
60274-60-4 | |
75°C to 77°C (4 mmHg) | |
C5H10ClI | |
MFCD00039416 | |
1732342 | |
pentane, 1-chloro-5-iodo, pentamethylene chloroiodide, 5-chloropentyl iodide, 5-iodopentyl chloride, l-chloro-5-iodopentane, 1-iodo-5-chloropentane, 1-chloro-5-iodo-pentane, pentane,1-chloro-5-iodo, acmc-1b36d, 1-chloro-5-iodopentane | |
GUUHKOCYWBEGGX-UHFFFAOYSA-N | |
1-chloro-5-iodopentane | |
108914 | |
97% |
Chemical Identifiers
60274-60-4 | |
232.489 | |
GUUHKOCYWBEGGX-UHFFFAOYSA-N | |
1-chloro-5-iodopentane |
C5H10ClI | |
MFCD00039416 | |
108914 | |
C(CCCl)CCI |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 262-137-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only