Learn More
1-Chloro-5-iodopentane, 97%, Thermo Scientific Chemicals
$166.11 - $166.11
Chemical Identifiers
CAS | 60274-60-4 |
---|---|
Molecular Formula | C5H10ClI |
Molecular Weight (g/mol) | 232.489 |
MDL Number | MFCD00039416 |
InChI Key | GUUHKOCYWBEGGX-UHFFFAOYSA-N |
PubChem CID | 108914 |
IUPAC Name | 1-chloro-5-iodopentane |
SMILES | C(CCCl)CCI |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0446909
|
Thermo Scientific Chemicals
L0446909 |
10 g |
Each for $166.11
|
|
|||||
Description
1-Chloro-5-iodopentane is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Chloro-5-iodopentane is used as a pharmaceutical intermediate.
Solubility
Not miscible or difficult to mix with water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to light.
Chemical Identifiers
60274-60-4 | |
232.489 | |
GUUHKOCYWBEGGX-UHFFFAOYSA-N | |
1-chloro-5-iodopentane |
C5H10ClI | |
MFCD00039416 | |
108914 | |
C(CCCl)CCI |
Specifications
60274-60-4 | |
75°C to 77°C (4 mmHg) | |
C5H10ClI | |
MFCD00039416 | |
1732342 | |
pentane, 1-chloro-5-iodo, pentamethylene chloroiodide, 5-chloropentyl iodide, 5-iodopentyl chloride, l-chloro-5-iodopentane, 1-iodo-5-chloropentane, 1-chloro-5-iodo-pentane, pentane,1-chloro-5-iodo, acmc-1b36d, 1-chloro-5-iodopentane | |
GUUHKOCYWBEGGX-UHFFFAOYSA-N | |
1-chloro-5-iodopentane | |
108914 | |
97% |
1.663 | |
>110°C (230°F) | |
1.5305 | |
10 g | |
Light sensitive | |
Not miscible or difficult to mix with water. | |
C(CCCl)CCI | |
232.489 | |
232.5 | |
1-Chloro-5-iodopentane |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 262-137-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only