Learn More
1-Chloro-6-iodohexane, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L0519809
Description
1-Chloro-6-iodohexane is used as :Pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Chloro-6-iodohexane is used as :Pharmaceutical intermediates.
Solubility
Not miscible in water.
Notes
Store at -20°C. Light Sensitive. Store away from oxidizing agents. Incompatible materials are Strong oxidizing agents.
Specifications
1-Chloro-6-iodohexane | |
C6H12ClI | |
MFCD00039417 | |
1732408 | |
hexane, 1-chloro-6-iodo, hexamethylene chloroiodide, 6-chlorohexyl iodide, acmc-20aovr, 6-chloro-1-iodohexane, 1-iodo-6-chloro hexane, 1-chloro-6-iodohexane, 1-chloro-6-iodohexane, stab. with copper chip, stabilized with copper chip | |
QTJHNJCILMMRIQ-UHFFFAOYSA-N | |
1-chloro-6-iodohexane | |
118713 | |
97% |
34683-73-3 | |
1.525 | |
10 g | |
Light sensitive | |
Not miscible in water. | |
C(CCCI)CCCl | |
246.516 | |
246.52 |
Chemical Identifiers
34683-73-3 | |
246.516 | |
QTJHNJCILMMRIQ-UHFFFAOYSA-N | |
118713 | |
C(CCCI)CCCl |
C6H12ClI | |
MFCD00039417 | |
hexane, 1-chloro-6-iodo, hexamethylene chloroiodide, 6-chlorohexyl iodide, acmc-20aovr, 6-chloro-1-iodohexane, 1-iodo-6-chloro hexane, 1-chloro-6-iodohexane, 1-chloro-6-iodohexane, stab. with copper chip, stabilized with copper chip | |
1-chloro-6-iodohexane |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
EINECSNumber : 252-144-0
TSCA : Yes
Recommended Storage : Store at -20°C
RUO – Research Use Only