missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Chloro-6-iodohexane, 97%, Thermo Scientific Chemicals

$239.23 - $790.74

Chemical Identifiers

CAS 34683-73-3
Molecular Formula C6H12ClI
Molecular Weight (g/mol) 246.516
MDL Number MFCD00039417
InChI Key QTJHNJCILMMRIQ-UHFFFAOYSA-N
Synonym hexane, 1-chloro-6-iodo, hexamethylene chloroiodide, 6-chlorohexyl iodide, acmc-20aovr, 6-chloro-1-iodohexane, 1-iodo-6-chloro hexane, 1-chloro-6-iodohexane, 1-chloro-6-iodohexane, stab. with copper chip, stabilized with copper chip
PubChem CID 118713
IUPAC Name 1-chloro-6-iodohexane
SMILES C(CCCI)CCCl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0519809
View Documents
Thermo Scientific Chemicals
L0519809
10 g
Each for $239.23
 
AAL0519818
View Documents
Thermo Scientific Chemicals
L0519818
50 g
Each for $790.74
 
Description

Description

1-Chloro-6-iodohexane is used as :Pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-6-iodohexane is used as :Pharmaceutical intermediates.

Solubility
Not miscible in water.

Notes
Store at -20°C. Light Sensitive. Store away from oxidizing agents. Incompatible materials are Strong oxidizing agents.
Specifications

Chemical Identifiers

34683-73-3
246.516
QTJHNJCILMMRIQ-UHFFFAOYSA-N
118713
C(CCCI)CCCl
C6H12ClI
MFCD00039417
hexane, 1-chloro-6-iodo, hexamethylene chloroiodide, 6-chlorohexyl iodide, acmc-20aovr, 6-chloro-1-iodohexane, 1-iodo-6-chloro hexane, 1-chloro-6-iodohexane, 1-chloro-6-iodohexane, stab. with copper chip, stabilized with copper chip
1-chloro-6-iodohexane

Specifications

34683-73-3
1.525
10 g
Light sensitive
Not miscible in water.
C(CCCI)CCCl
246.516
246.52
1-Chloro-6-iodohexane
C6H12ClI
MFCD00039417
1732408
hexane, 1-chloro-6-iodo, hexamethylene chloroiodide, 6-chlorohexyl iodide, acmc-20aovr, 6-chloro-1-iodohexane, 1-iodo-6-chloro hexane, 1-chloro-6-iodohexane, 1-chloro-6-iodohexane, stab. with copper chip, stabilized with copper chip
QTJHNJCILMMRIQ-UHFFFAOYSA-N
1-chloro-6-iodohexane
118713
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

EINECSNumber : 252-144-0

TSCA : Yes

Recommended Storage : Store at -20°C

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only