missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Chloro-6-iodohexane, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L0519818

 View more versions of this product

Catalog No. AAL0519818


Explore available promotions

Description

Description

1-Chloro-6-iodohexane is used as :Pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-6-iodohexane is used as :Pharmaceutical intermediates.

Solubility
Not miscible in water.

Notes
Store at -20°C. Light Sensitive. Store away from oxidizing agents. Incompatible materials are Strong oxidizing agents.
Specifications

Specifications

1-Chloro-6-iodohexane
C6H12ClI
MFCD00039417
1732408
hexane, 1-chloro-6-iodo, hexamethylene chloroiodide, 6-chlorohexyl iodide, acmc-20aovr, 6-chloro-1-iodohexane, 1-iodo-6-chloro hexane, 1-chloro-6-iodohexane, 1-chloro-6-iodohexane, stab. with copper chip, stabilized with copper chip
QTJHNJCILMMRIQ-UHFFFAOYSA-N
1-chloro-6-iodohexane
118713
97%
34683-73-3
1.525
50 g
Light sensitive
Not miscible in water.
C(CCCI)CCCl
246.516
246.52

Chemical Identifiers

34683-73-3
246.516
QTJHNJCILMMRIQ-UHFFFAOYSA-N
118713
C(CCCI)CCCl
C6H12ClI
MFCD00039417
hexane, 1-chloro-6-iodo, hexamethylene chloroiodide, 6-chlorohexyl iodide, acmc-20aovr, 6-chloro-1-iodohexane, 1-iodo-6-chloro hexane, 1-chloro-6-iodohexane, 1-chloro-6-iodohexane, stab. with copper chip, stabilized with copper chip
1-chloro-6-iodohexane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

EINECSNumber : 252-144-0

TSCA : Yes

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only