missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Chloro-N,N,2-trimethylpropenylamine, 98.5+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 300270050

 View more versions of this product

Catalog No. AC300270050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Chloro-N, N, 2-trimethylpropenylamine
Colorless to Yellow
129.0°C to 130.0°C
Authentic
Glass bottle
1.4520 to 1.4560
1.01
Solubility in water: .. Other solubilities: soluble in most organic solvents
CC(=C(N(C)C)Cl)C
133.62
133.62
Liquid
26189-59-3
1.0100g/mL
27°C
98.5% min. (GC)
C6H12ClN
5 mL
1-chloro-n,n,2-trimethylpropenylamine, 1-chloro-n,n,2-trimethyl-1-propenylamine, 1-chloro-n,n,2-trimethyl-1-propen-1-amine, ghosez inverted exclamation markas reagent, 1-propen-1-amine, 1-chloro-n,n,2-trimethyl, 1-chloro-2-methylprop-1-en-1-yl dimethylamine, acmc-1ceik, 1-chloro-1-dimethylaminoisobutene, 1-chloro-2,n,n-trimethylpropenylamine, 1-chloro-n,n-2-trimethylpropenylamine
GQIRIWDEZSKOCN-UHFFFAOYSA-N
1-chloro-N,N,2-trimethylprop-1-en-1-amine
640792
98.5+%

Chemical Identifiers

26189-59-3
133.62
1-chloro-n,n,2-trimethylpropenylamine, 1-chloro-n,n,2-trimethyl-1-propenylamine, 1-chloro-n,n,2-trimethyl-1-propen-1-amine, ghosez inverted exclamation markas reagent, 1-propen-1-amine, 1-chloro-n,n,2-trimethyl, 1-chloro-2-methylprop-1-en-1-yl dimethylamine, acmc-1ceik, 1-chloro-1-dimethylaminoisobutene, 1-chloro-2,n,n-trimethylpropenylamine, 1-chloro-n,n-2-trimethylpropenylamine
1-chloro-N,N,2-trimethylprop-1-en-1-amine
C6H12ClN
GQIRIWDEZSKOCN-UHFFFAOYSA-N
640792
CC(=C(N(C)C)Cl)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapour.
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only