missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Chloro-N,N,2-trimethylpropenylamine, 98.5+%, Thermo Scientific Chemicals

$266.43 - $1065.35

Chemical Identifiers

CAS 26189-59-3
Molecular Formula C6H12ClN
Molecular Weight (g/mol) 133.62
InChI Key GQIRIWDEZSKOCN-UHFFFAOYSA-N
Synonym 1-chloro-n,n,2-trimethylpropenylamine, 1-chloro-n,n,2-trimethyl-1-propenylamine, 1-chloro-n,n,2-trimethyl-1-propen-1-amine, ghosez inverted exclamation markas reagent, 1-propen-1-amine, 1-chloro-n,n,2-trimethyl, 1-chloro-2-methylprop-1-en-1-yl dimethylamine, acmc-1ceik, 1-chloro-1-dimethylaminoisobutene, 1-chloro-2,n,n-trimethylpropenylamine, 1-chloro-n,n-2-trimethylpropenylamine
PubChem CID 640792
IUPAC Name 1-chloro-N,N,2-trimethylprop-1-en-1-amine
SMILES CC(=C(N(C)C)Cl)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC300270050
SDS View Documents
Thermo Scientific Chemicals
300270050
5 mL Glass bottle
Each for $266.43
 
AC300270250
SDS View Documents
Thermo Scientific Chemicals
300270250
25 mL Glass bottle
Each for $1,065.35
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

26189-59-3
133.62
1-chloro-n,n,2-trimethylpropenylamine, 1-chloro-n,n,2-trimethyl-1-propenylamine, 1-chloro-n,n,2-trimethyl-1-propen-1-amine, ghosez inverted exclamation markas reagent, 1-propen-1-amine, 1-chloro-n,n,2-trimethyl, 1-chloro-2-methylprop-1-en-1-yl dimethylamine, acmc-1ceik, 1-chloro-1-dimethylaminoisobutene, 1-chloro-2,n,n-trimethylpropenylamine, 1-chloro-n,n-2-trimethylpropenylamine
1-chloro-N,N,2-trimethylprop-1-en-1-amine
C6H12ClN
GQIRIWDEZSKOCN-UHFFFAOYSA-N
640792
CC(=C(N(C)C)Cl)C

Specifications

26189-59-3
1.0100g/mL
27°C
98.5% min. (GC)
C6H12ClN
5 mL
1-chloro-n,n,2-trimethylpropenylamine, 1-chloro-n,n,2-trimethyl-1-propenylamine, 1-chloro-n,n,2-trimethyl-1-propen-1-amine, ghosez inverted exclamation markas reagent, 1-propen-1-amine, 1-chloro-n,n,2-trimethyl, 1-chloro-2-methylprop-1-en-1-yl dimethylamine, acmc-1ceik, 1-chloro-1-dimethylaminoisobutene, 1-chloro-2,n,n-trimethylpropenylamine, 1-chloro-n,n-2-trimethylpropenylamine
GQIRIWDEZSKOCN-UHFFFAOYSA-N
1-chloro-N,N,2-trimethylprop-1-en-1-amine
640792
98.5+%
1-Chloro-N, N, 2-trimethylpropenylamine
Colorless to Yellow
129.0°C to 130.0°C
Authentic
Glass bottle
1.4520 to 1.4560
1.01
Solubility in water: .. Other solubilities: soluble in most organic solvents
CC(=C(N(C)C)Cl)C
133.62
133.62
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapour.
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only