missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Dodecanethiol, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 117620025

 View more versions of this product

Catalog No. AC117620025


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Dodecanethiol
97.5
-7.0°C
266.0°C to 283.0°C
Authentic
Glass bottle
1.4580 to 1.4600
MFCD00004885
16,356
1-dodecanethiol, dodecyl mercaptan, dodecanethiol, n-dodecanethiol, lauryl mercaptan, n-dodecyl mercaptan, n-dodecylmercaptan, 1-mercaptododecane, n-lauryl mercaptan, 1-dodecyl mercaptan
WNAHIZMDSQCWRP-UHFFFAOYSA-N
dodecane-1-thiol
8195
98%
112-55-0
100.0
0.8400g/mL
128°C
97.5% min. (GC)
C12H26S
CH3(CH2)11SH
2.5 L
0.84
Solubility in water: insoluble. Other solubilities: soluble in hydrocarbons, slightly soluble in alcohol
CCCCCCCCCCCCS
202.40
202.4
Liquid

Chemical Identifiers

112-55-0
202.40
WNAHIZMDSQCWRP-UHFFFAOYSA-N
8195
CCCCCCCCCCCCS
C12H26S
MFCD00004885
1-dodecanethiol, dodecyl mercaptan, dodecanethiol, n-dodecanethiol, lauryl mercaptan, n-dodecyl mercaptan, n-dodecylmercaptan, 1-mercaptododecane, n-lauryl mercaptan, 1-dodecyl mercaptan
dodecane-1-thiol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

EINECSNumber : 203-984-1

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only