Learn More
1-Dodecanethiol, 98%, Thermo Scientific Chemicals
$86.11 - $196.61
Chemical Identifiers
CAS | 112-55-0 |
---|---|
Molecular Formula | C12H26S |
Molecular Weight (g/mol) | 202.40 |
MDL Number | MFCD00004885 |
InChI Key | WNAHIZMDSQCWRP-UHFFFAOYSA-N |
Synonym | 1-dodecanethiol, dodecyl mercaptan, dodecanethiol, n-dodecanethiol, lauryl mercaptan, n-dodecyl mercaptan, n-dodecylmercaptan, 1-mercaptododecane, n-lauryl mercaptan, 1-dodecyl mercaptan |
PubChem CID | 8195 |
IUPAC Name | dodecane-1-thiol |
SMILES | CCCCCCCCCCCCS |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC117625000
|
Thermo Scientific Chemicals
117625000 |
500 mL | Glass bottle |
Each for $86.11
|
|
||||
AC117620025
|
Thermo Scientific Chemicals
117620025 |
2.5 L | Glass bottle |
Each for $196.61
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
112-55-0 | |
202.40 | |
WNAHIZMDSQCWRP-UHFFFAOYSA-N | |
8195 | |
CCCCCCCCCCCCS |
C12H26S | |
MFCD00004885 | |
1-dodecanethiol, dodecyl mercaptan, dodecanethiol, n-dodecanethiol, lauryl mercaptan, n-dodecyl mercaptan, n-dodecylmercaptan, 1-mercaptododecane, n-lauryl mercaptan, 1-dodecyl mercaptan | |
dodecane-1-thiol |
Specifications
112-55-0 | |
100.0 | |
0.8400g/mL | |
128°C | |
97.5% min. (GC) | |
C12H26S | |
CH3(CH2)11SH | |
500 mL | |
0.84 | |
Solubility in water: insoluble. Other solubilities: soluble in hydrocarbons, slightly soluble in alcohol | |
CCCCCCCCCCCCS | |
202.40 | |
202.4 | |
Liquid |
97.5 | |
-7.0°C | |
266.0°C to 283.0°C | |
Authentic | |
Glass bottle | |
1.4580 to 1.4600 | |
MFCD00004885 | |
16,356 | |
1-dodecanethiol, dodecyl mercaptan, dodecanethiol, n-dodecanethiol, lauryl mercaptan, n-dodecyl mercaptan, n-dodecylmercaptan, 1-mercaptododecane, n-lauryl mercaptan, 1-dodecyl mercaptan | |
WNAHIZMDSQCWRP-UHFFFAOYSA-N | |
dodecane-1-thiol | |
8195 | |
98% | |
1-Dodecanethiol |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
EINECSNumber : 203-984-1
RUO – Research Use Only