Learn More
1-Dodecanol, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 155450100
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1-Dodecanol | |
97.5 | |
22.0°C to 27.0°C | |
0.8350g/mL | |
119°C | |
97.5% min. (GC) | |
C12H26O | |
MFCD00004753 | |
01, 428 | |
15, 3451 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether | |
CCCCCCCCCCCCO | |
186.34 | |
10 mPa.s (40°C) | |
186.34 | |
98% |
112-53-8 | |
100.0 | |
Colorless to Yellow | |
258.0°C to 265.0°C | |
Authentic | |
Plastic drum | |
CH3(CH2)11OH | |
10 L | |
0.835 | |
1-dodecanol, dodecyl alcohol, lauryl alcohol, dodecanol, n-dodecyl alcohol, lauric alcohol, laurinic alcohol, undecyl carbinol, dodecylalcohol, 1-dodecyl alcohol | |
LQZZUXJYWNFBMV-UHFFFAOYSA-N | |
dodecan-1-ol | |
8193 | |
CHEBI:28878 | |
<0.1mbar at 20°C | |
Liquid |
Chemical Identifiers
112-53-8 | |
186.34 | |
LQZZUXJYWNFBMV-UHFFFAOYSA-N | |
8193 | |
dodecan-1-ol |
C12H26O | |
MFCD00004753 | |
1-dodecanol, dodecyl alcohol, lauryl alcohol, dodecanol, n-dodecyl alcohol, lauric alcohol, laurinic alcohol, undecyl carbinol, dodecylalcohol, 1-dodecyl alcohol | |
CHEBI:28878 | |
CCCCCCCCCCCCO |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Very toxic to aquatic life.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protec
GHS Signal Word: Warning
EINECSNumber : 203-982-
RUO – Research Use Only