Learn More
1-Dodecanol, 98%, Thermo Scientific Chemicals
$71.08 - $484.86
Chemical Identifiers
CAS | 112-53-8 |
---|---|
Molecular Formula | C12H26O |
Molecular Weight (g/mol) | 186.34 |
MDL Number | MFCD00004753 |
InChI Key | LQZZUXJYWNFBMV-UHFFFAOYSA-N |
Synonym | 1-dodecanol, dodecyl alcohol, lauryl alcohol, dodecanol, n-dodecyl alcohol, lauric alcohol, laurinic alcohol, undecyl carbinol, dodecylalcohol, 1-dodecyl alcohol |
PubChem CID | 8193 |
ChEBI | CHEBI:28878 |
IUPAC Name | dodecan-1-ol |
SMILES | CCCCCCCCCCCCO |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC155455000
|
Thermo Scientific Chemicals
155455000 |
500 mL |
Each for $71.08
|
|
|||||
AC155450010
|
Thermo Scientific Chemicals
155450010 |
1 L |
Each for $105.86
|
|
|||||
AC155450100
|
Thermo Scientific Chemicals
155450100 |
10 L |
Each for $484.86
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
112-53-8 | |
186.34 | |
LQZZUXJYWNFBMV-UHFFFAOYSA-N | |
8193 | |
dodecan-1-ol |
C12H26O | |
MFCD00004753 | |
1-dodecanol, dodecyl alcohol, lauryl alcohol, dodecanol, n-dodecyl alcohol, lauric alcohol, laurinic alcohol, undecyl carbinol, dodecylalcohol, 1-dodecyl alcohol | |
CHEBI:28878 | |
CCCCCCCCCCCCO |
Specifications
112-53-8 | |
100.0 | |
Colorless to Yellow | |
258.0°C to 265.0°C | |
Authentic | |
Glass Bottle | |
CH3(CH2)11OH | |
500 mL | |
15, 3451 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether | |
CCCCCCCCCCCCO | |
186.34 | |
CHEBI:28878 | |
<0.1mbar at 20°C | |
Liquid |
97.5 | |
22.0°C to 27.0°C | |
0.8350g/mL | |
119°C | |
98% | |
C12H26O | |
MFCD00004753 | |
01, 428 | |
1-dodecanol, dodecyl alcohol, lauryl alcohol, dodecanol, n-dodecyl alcohol, lauric alcohol, laurinic alcohol, undecyl carbinol, dodecylalcohol, 1-dodecyl alcohol | |
LQZZUXJYWNFBMV-UHFFFAOYSA-N | |
dodecan-1-ol | |
8193 | |
186.34 | |
98% | |
1-Dodecanol |
Safety and Handling
GHS P Statement
Causes serious eye irritation.
Very toxic to aquatic life.
GHS P Statement
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
RUO – Research Use Only