missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Dodecene, 93-95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 117631000

 View more versions of this product

Catalog No. AC117631000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Dodecene
112-41-4
100.0
0.7500g/mL
77°C
Glass bottle
1.4284 to 1.4304
MFCD00008961
01,225
1-dodecene, dodecene, adacene 12, n-dodec-1-ene, dodecylene, alpha-dodecene, alpha-dodecylene, dodecene-1, .alpha.-dodecene, unii-wye669f3gr
CRSBERNSMYQZNG-UHFFFAOYSA-N
dodec-1-ene
8183
93 to 95%
93-95%
93.0
-35.0°C
213.0°C
93% min. (GC)
C12H24
CH3(CH2)9CH=CH2
100 g
0.75
Solubility in water: insoluble. Other solubilities: soluble in alcohol, acetone and ether
CCCCCCCCCCC=C
168.32
168.32

Chemical Identifiers

112-41-4
168.32
CRSBERNSMYQZNG-UHFFFAOYSA-N
8183
CCCCCCCCCCC=C
C12H24
MFCD00008961
1-dodecene, dodecene, adacene 12, n-dodec-1-ene, dodecylene, alpha-dodecene, alpha-dodecylene, dodecene-1, .alpha.-dodecene, unii-wye669f3gr
dodec-1-ene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
May be fatal if swallowed and enters airways.
Very toxic to aquatic life with long lasting effects.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap an

GHS Signal Word: Danger

EINECSNumber : 203-968-4

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only