Learn More
1-Dodecene, 93-95%, Thermo Scientific Chemicals
$95.27 - $790.26
Chemical Identifiers
CAS | 112-41-4 |
---|---|
Molecular Formula | C12H24 |
Molecular Weight (g/mol) | 168.32 |
MDL Number | MFCD00008961 |
InChI Key | CRSBERNSMYQZNG-UHFFFAOYSA-N |
Synonym | 1-dodecene, dodecene, adacene 12, n-dodec-1-ene, dodecylene, alpha-dodecene, alpha-dodecylene, dodecene-1, .alpha.-dodecene, unii-wye669f3gr |
PubChem CID | 8183 |
IUPAC Name | dodec-1-ene |
SMILES | CCCCCCCCCCC=C |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC117631000
|
Thermo Scientific Chemicals
117631000 |
100 g | Glass bottle |
Each for $95.27
|
|
|||||
AC117635000
|
Thermo Scientific Chemicals
117635000 |
500 g | Glass bottle |
Each for $188.63
|
|
|||||
AC117630025
|
Thermo Scientific Chemicals
117630025 |
2.5 kg | Plastic drum |
Each for $790.26
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
112-41-4 | |
168.32 | |
CRSBERNSMYQZNG-UHFFFAOYSA-N | |
8183 | |
CCCCCCCCCCC=C |
C12H24 | |
MFCD00008961 | |
1-dodecene, dodecene, adacene 12, n-dodec-1-ene, dodecylene, alpha-dodecene, alpha-dodecylene, dodecene-1, .alpha.-dodecene, unii-wye669f3gr | |
dodec-1-ene |
Specifications
112-41-4 | |
100.0 | |
0.7500g/mL | |
77°C | |
Glass bottle | |
1.4284 to 1.4304 | |
MFCD00008961 | |
01,225 | |
1-dodecene, dodecene, adacene 12, n-dodec-1-ene, dodecylene, alpha-dodecene, alpha-dodecylene, dodecene-1, .alpha.-dodecene, unii-wye669f3gr | |
CRSBERNSMYQZNG-UHFFFAOYSA-N | |
dodec-1-ene | |
8183 | |
93 to 95% |
93.0 | |
-35.0°C | |
213.0°C | |
93% min. (GC) | |
C12H24 | |
CH3(CH2)9CH=CH2 | |
100 g | |
0.75 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol, acetone and ether | |
CCCCCCCCCCC=C | |
168.32 | |
168.32 | |
1-Dodecene, 93-95% |
Safety and Handling
GHS H Statement
May be fatal if swallowed and enters airways.
Very toxic to aquatic life with long lasting effects.
Repeated exposure may cause skin dryness or cracking.
GHS P Statement
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap an
GHS Signal Word: Danger
EINECSNumber : 203-968-4
TSCA : TSCA
RUO – Research Use Only