Learn More
1-Dodecene, 93-95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 117635000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1-Dodecene | |
112-41-4 | |
100.0 | |
0.7500g/mL | |
77°C | |
Glass bottle | |
1.4284 to 1.4304 | |
MFCD00008961 | |
01,225 | |
1-dodecene, dodecene, adacene 12, n-dodec-1-ene, dodecylene, alpha-dodecene, alpha-dodecylene, dodecene-1, .alpha.-dodecene, unii-wye669f3gr | |
CRSBERNSMYQZNG-UHFFFAOYSA-N | |
dodec-1-ene | |
8183 | |
93 to 95% |
93-95% | |
93.0 | |
-35.0°C | |
213.0°C | |
93% min. (GC) | |
C12H24 | |
CH3(CH2)9CH=CH2 | |
500 g | |
0.75 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol, acetone and ether | |
CCCCCCCCCCC=C | |
168.32 | |
168.32 |
Chemical Identifiers
112-41-4 | |
168.32 | |
CRSBERNSMYQZNG-UHFFFAOYSA-N | |
8183 | |
CCCCCCCCCCC=C |
C12H24 | |
MFCD00008961 | |
1-dodecene, dodecene, adacene 12, n-dodec-1-ene, dodecylene, alpha-dodecene, alpha-dodecylene, dodecene-1, .alpha.-dodecene, unii-wye669f3gr | |
dodec-1-ene |
Safety and Handling
GHS H Statement
May be fatal if swallowed and enters airways.
Very toxic to aquatic life with long lasting effects.
Repeated exposure may cause skin dryness or cracking.
GHS P Statement
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap an
GHS Signal Word: Danger
EINECSNumber : 203-968-4
TSCA : TSCA
RUO – Research Use Only