Learn More
1-Ethyl-3-methylimidazolium ethyl sulfate, 99%, Thermo Scientific Chemicals
$186.27 - $821.78
Chemical Identifiers
CAS | 342573-75-5 |
---|---|
Molecular Formula | C8H16N2O4S |
Molecular Weight (g/mol) | 236.286 |
MDL Number | MFCD06798189 |
InChI Key | VRFOKYHDLYBVAL-UHFFFAOYSA-M |
Synonym | 1-ethyl-3-methylimidazolium ethyl sulfate, 1-ethyl-3-methylimidazolium ethylsulfate, 1-ethyl-3-methyl-1h-imidazol-3-ium ethyl sulfate, basionics™ lq 01, dsstox_cid_29130, dsstox_rid_83349, dsstox_gsid_49274, ksc221o0f, 1h-imidazolium, 3-ethyl-1-methyl-, ethyl sulfate 1:1, 1-ethyl-3-me-thylimidazolium ethylsulfate |
PubChem CID | 12095229 |
IUPAC Name | 1-ethyl-3-methylimidazol-3-ium;ethyl sulfate |
SMILES | CCN1C=C[N+](=C1)C.CCOS(=O)(=O)[O-] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH2757118
|
Thermo Scientific Chemicals
H2757118 |
50 g |
Each for $186.27
|
|
|||||
AAH2757130
|
Thermo Scientific Chemicals
H2757130 |
250 g |
Each for $821.78
|
|
|||||
Description
1-Ethyl-3-methylimidazolium ethyl sulfate is an ionic liquid having free form chloride used as an alternative to the corresponding halide salts in metathesis reactions to prepare other ionic liquids including 1-butyl-3-methylimidazolium hexafluorophosphate. It is also utilized in the study of layering and shear properties of ionic liquid between nano-films and mica surfaces.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Ethyl-3-methylimidazolium ethyl sulfate is an ionic liquid having free form chloride used as an alternative to the corresponding halide salts in metathesis reactions to prepare other ionic liquids including 1-butyl-3-methylimidazolium hexafluorophosphate. It is also utilized in the study of layering and shear properties of ionic liquid between nano-films and mica surfaces.
Solubility
Miscible with water and alcohol.
Notes
Hygroscopic. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and bases.
Chemical Identifiers
342573-75-5 | |
236.286 | |
VRFOKYHDLYBVAL-UHFFFAOYSA-M | |
12095229 | |
CCN1C=C[N+](=C1)C.CCOS(=O)(=O)[O-] |
C8H16N2O4S | |
MFCD06798189 | |
1-ethyl-3-methylimidazolium ethyl sulfate, 1-ethyl-3-methylimidazolium ethylsulfate, 1-ethyl-3-methyl-1h-imidazol-3-ium ethyl sulfate, basionics™ lq 01, dsstox_cid_29130, dsstox_rid_83349, dsstox_gsid_49274, ksc221o0f, 1h-imidazolium, 3-ethyl-1-methyl-, ethyl sulfate 1:1, 1-ethyl-3-me-thylimidazolium ethylsulfate | |
1-ethyl-3-methylimidazol-3-ium;ethyl sulfate |
Specifications
342573-75-5 | |
1.24 | |
C8H16N2O4S | |
50 g | |
Miscible with water and alcohol. | |
CCN1C=C[N+](=C1)C.CCOS(=O)(=O)[O-] | |
236.286 | |
236.29 | |
1-Ethyl-3-methylimidazolium ethyl sulfate |
<-30°C | |
162°C (324°F) | |
MFCD06798189 | |
1-ethyl-3-methylimidazolium ethyl sulfate, 1-ethyl-3-methylimidazolium ethylsulfate, 1-ethyl-3-methyl-1h-imidazol-3-ium ethyl sulfate, basionics™ lq 01, dsstox_cid_29130, dsstox_rid_83349, dsstox_gsid_49274, ksc221o0f, 1h-imidazolium, 3-ethyl-1-methyl-, ethyl sulfate 1:1, 1-ethyl-3-me-thylimidazolium ethylsulfate | |
VRFOKYHDLYBVAL-UHFFFAOYSA-M | |
1-ethyl-3-methylimidazol-3-ium;ethyl sulfate | |
12095229 | |
99% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only