missing translation for 'onlineSavingsMsg'
Learn More

1-Ethylcyclohexanol, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 453350250

 Afficher plus de versions de ce produit

Numéro de catalogue. AC453350250

Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Découvrez plus d'offres spéciales

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Spécifications

Spécifications

1-Ethylcyclohexanol
0.9190g/mL
68°C
C8H16O
25mL
1-ethylcyclohexanol, cyclohexanol, 1-ethyl, unii-1ves5dbc5c, 1-aethyl-cyclohexanol-1, 1-ethyl-1-cyclohexanol, 1ves5dbc5c, 1-aethyl-cyclohexanol-1 german, ethylcyclohexanol, cyclohexanol, ethyl, 1-ethyl-cyclohexan-1-ol
CCC1(O)CCCCC1
128.22
128.22
1940-18-7
166.0°C
96% min. (GC)
MFCD00021402
0.919
BUCJHJXFXUZJHL-UHFFFAOYSA-N
1-ethylcyclohexan-1-ol
16021
97%

Identifiants chimiques

1940-18-7
128.22
BUCJHJXFXUZJHL-UHFFFAOYSA-N
16021
CCC1(O)CCCCC1
C8H16O
MFCD00021402
1-ethylcyclohexanol, cyclohexanol, 1-ethyl, unii-1ves5dbc5c, 1-aethyl-cyclohexanol-1, 1-ethyl-1-cyclohexanol, 1ves5dbc5c, 1-aethyl-cyclohexanol-1 german, ethylcyclohexanol, cyclohexanol, ethyl, 1-ethyl-cyclohexan-1-ol
1-ethylcyclohexan-1-ol
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.

GHS Signal Word: Warning

FDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only