missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Fluoronaphthalene 98.0+%, TCI America™
Supplier: TCI America F021225G
Specifications
1-Fluoronaphthalene | |
Yellow | |
C10H7F | |
25 g | |
CWLKTJOTWITYSI-UHFFFAOYSA-N | |
1-fluoronaphthalene | |
9450 | |
≥98.0% (GC) |
321-38-0 | |
212°C | |
MFCD00003873 | |
fluoronaphthalene, naphthalene, 1-fluoro, alpha-fluoronaphthalene, 1-fluornaftalen, .alpha.-fluoronaphthalene, 1-fluornaftalen czech, 1-fluoro naphthalene, fluoronaphtalene, fluoronapthalene, 1-fluoronapthalene | |
C1=CC=C2C(=C1)C=CC=C2F | |
146.164 | |
146.16 | |
Liquid |
Chemical Identifiers
321-38-0 | |
146.164 | |
CWLKTJOTWITYSI-UHFFFAOYSA-N | |
9450 | |
C1=CC=C2C(=C1)C=CC=C2F |
C10H7F | |
MFCD00003873 | |
fluoronaphthalene, naphthalene, 1-fluoro, alpha-fluoronaphthalene, 1-fluornaftalen, .alpha.-fluoronaphthalene, 1-fluornaftalen czech, 1-fluoro naphthalene, fluoronaphtalene, fluoronapthalene, 1-fluoronapthalene | |
1-fluoronaphthalene |
Safety and Handling
RTECSNumber : QJ7100000
TSCA : Yes