missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Fluoronaphthalene 98.0+%, TCI America™
$37.52 - $88.87
Chemical Identifiers
CAS | 321-38-0 |
---|---|
Molecular Formula | C10H7F |
Molecular Weight (g/mol) | 146.164 |
MDL Number | MFCD00003873 |
InChI Key | CWLKTJOTWITYSI-UHFFFAOYSA-N |
Synonym | fluoronaphthalene, naphthalene, 1-fluoro, alpha-fluoronaphthalene, 1-fluornaftalen, .alpha.-fluoronaphthalene, 1-fluornaftalen czech, 1-fluoro naphthalene, fluoronaphtalene, fluoronapthalene, 1-fluoronapthalene |
PubChem CID | 9450 |
IUPAC Name | 1-fluoronaphthalene |
SMILES | C1=CC=C2C(=C1)C=CC=C2F |
Chemical Identifiers
321-38-0 | |
146.164 | |
CWLKTJOTWITYSI-UHFFFAOYSA-N | |
9450 | |
C1=CC=C2C(=C1)C=CC=C2F |
C10H7F | |
MFCD00003873 | |
fluoronaphthalene, naphthalene, 1-fluoro, alpha-fluoronaphthalene, 1-fluornaftalen, .alpha.-fluoronaphthalene, 1-fluornaftalen czech, 1-fluoro naphthalene, fluoronaphtalene, fluoronapthalene, 1-fluoronapthalene | |
1-fluoronaphthalene |
Specifications
321-38-0 | |
212°C | |
MFCD00003873 | |
fluoronaphthalene, naphthalene, 1-fluoro, alpha-fluoronaphthalene, 1-fluornaftalen, .alpha.-fluoronaphthalene, 1-fluornaftalen czech, 1-fluoro naphthalene, fluoronaphtalene, fluoronapthalene, 1-fluoronapthalene | |
C1=CC=C2C(=C1)C=CC=C2F | |
146.164 | |
146.16 | |
Liquid |
Yellow | |
C10H7F | |
5 g | |
CWLKTJOTWITYSI-UHFFFAOYSA-N | |
1-fluoronaphthalene | |
9450 | |
≥98.0% (GC) | |
1-Fluoronaphthalene |
Safety and Handling
RTECSNumber : QJ7100000
TSCA : Yes