Learn More
1-Heptanethiol, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L0707922
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1-Heptanethiol | |
-43°C | |
173°C to 175°C | |
C7H16S | |
MFCD00004911 | |
UN3336 | |
Air Sensitive | |
VPIAKHNXCOTPAY-UHFFFAOYSA-N | |
heptane-1-thiol | |
15422 | |
98% |
1639-09-4 | |
0.854 | |
46°C (114°F) | |
1.4515 | |
100 g | |
1731687 | |
1-heptanethiol, heptyl mercaptan, n-heptylmercaptan, heptyl thiol, heptylthiol, 1-heptylthiol, n-heptyl mercaptan, usaf ek-2122, unii-380ze61gr2, normal-heptyl mercaptan | |
CCCCCCCS | |
132.265 | |
132.27 |
Chemical Identifiers
1639-09-4 | |
132.265 | |
VPIAKHNXCOTPAY-UHFFFAOYSA-N | |
15422 | |
CCCCCCCS |
C7H16S | |
MFCD00004911 | |
1-heptanethiol, heptyl mercaptan, n-heptylmercaptan, heptyl thiol, heptylthiol, 1-heptylthiol, n-heptyl mercaptan, usaf ek-2122, unii-380ze61gr2, normal-heptyl mercaptan | |
heptane-1-thiol |
Safety and Handling
GHS H Statement
H226-H335
Flammable liquid and vapor.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c
H226-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: MERCAPTANS, LIQUID, FLAMMABLE, N.O.S.
EINECSNumber : 216-678-8
RTECSNumber : MJ1400000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only