missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Heptanethiol, 98%, Thermo Scientific Chemicals

$96.96 - $242.88

Chemical Identifiers

CAS 1639-09-4
Molecular Formula C7H16S
Molecular Weight (g/mol) 132.265
MDL Number MFCD00004911
InChI Key VPIAKHNXCOTPAY-UHFFFAOYSA-N
Synonym 1-heptanethiol, heptyl mercaptan, n-heptylmercaptan, heptyl thiol, heptylthiol, 1-heptylthiol, n-heptyl mercaptan, usaf ek-2122, unii-380ze61gr2, normal-heptyl mercaptan
PubChem CID 15422
IUPAC Name heptane-1-thiol
SMILES CCCCCCCS
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0707914
View Documents
Thermo Scientific Chemicals
L0707914
25 g
Each for $96.96
 
AAL0707922
View Documents
Thermo Scientific Chemicals
L0707922
100 g
Each for $242.88
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1639-09-4
132.265
VPIAKHNXCOTPAY-UHFFFAOYSA-N
15422
CCCCCCCS
C7H16S
MFCD00004911
1-heptanethiol, heptyl mercaptan, n-heptylmercaptan, heptyl thiol, heptylthiol, 1-heptylthiol, n-heptyl mercaptan, usaf ek-2122, unii-380ze61gr2, normal-heptyl mercaptan
heptane-1-thiol

Specifications

1639-09-4
0.854
46°C (114°F)
1.4515
25 g
1731687
1-heptanethiol, heptyl mercaptan, n-heptylmercaptan, heptyl thiol, heptylthiol, 1-heptylthiol, n-heptyl mercaptan, usaf ek-2122, unii-380ze61gr2, normal-heptyl mercaptan
CCCCCCCS
132.265
132.27
1-Heptanethiol
-43°C
173°C to 175°C
C7H16S
MFCD00004911
UN3336
Air Sensitive
VPIAKHNXCOTPAY-UHFFFAOYSA-N
heptane-1-thiol
15422
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H335
Flammable liquid and vapor.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c

H226-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: MERCAPTANS, LIQUID, FLAMMABLE, N.O.S.

EINECSNumber : 216-678-8

RTECSNumber : MJ1400000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only