Learn More
1-Heptanol, 98%, Thermo Scientific Chemicals
$56.79 - $239.96
Chemical Identifiers
CAS | 111-70-6 |
---|---|
Molecular Formula | C7H16O |
Molecular Weight (g/mol) | 116.20 |
MDL Number | MFCD00002986 |
InChI Key | BBMCTIGTTCKYKF-UHFFFAOYSA-N |
Synonym | 1-heptanol, heptyl alcohol, heptanol, n-heptanol, n-heptyl alcohol, enanthic alcohol, gentanol, 1-hydroxyheptane, n-heptan-1-ol, enanthyl alcohol |
PubChem CID | 8129 |
IUPAC Name | heptan-1-ol |
SMILES | CCCCCCCO |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC120362500
|
Thermo Scientific Chemicals
120362500 |
250 mL | Glass bottle |
N/A
|
|
||||
AC120360010
|
Thermo Scientific Chemicals
120360010 |
1 L | Glass bottle |
N/A
|
|
||||
AC120360025
|
Thermo Scientific Chemicals
120360025 |
2.5 L | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
111-70-6 | |
116.20 | |
BBMCTIGTTCKYKF-UHFFFAOYSA-N | |
8129 | |
CCCCCCCO |
C7H16O | |
MFCD00002986 | |
1-heptanol, heptyl alcohol, heptanol, n-heptanol, n-heptyl alcohol, enanthic alcohol, gentanol, 1-hydroxyheptane, n-heptan-1-ol, enanthyl alcohol | |
heptan-1-ol |
Specifications
111-70-6 | |
97.5 | |
-35.0°C | |
176.0°C | |
Authentic | |
Glass bottle | |
1.4230 to 1.4250 | |
MFCD00002986 | |
01, 414 | |
0.8219 | |
1-heptanol, heptyl alcohol, heptanol, n-heptanol, n-heptyl alcohol, enanthic alcohol, gentanol, 1-hydroxyheptane, n-heptan-1-ol, enanthyl alcohol | |
BBMCTIGTTCKYKF-UHFFFAOYSA-N | |
heptan-1-ol | |
8129 | |
98% | |
Liquid |
0.5mg KOH/g max. | |
100.0 | |
0.8219g/mL | |
73°C | |
97.5% min. (GC) | |
C7H16O | |
CH3(CH2)6OH | |
250 mL | |
0.01% max. | |
15, 4696 | |
Solubility in water: 2.85g/L (100°C). Other solubilities: 5.15g/L water (130°C), soluble in ethanol, ether and usual organic, solvents | |
CCCCCCCO | |
116.20 | |
116.2 | |
Extra Pure | |
1-Heptanol |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Warning
EINECSNumber : 203-897-9
RUO – Research Use Only