missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Heptylamine, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1792230

 View more versions of this product

Catalog No. AAA1792230


Explore available promotions

Description

Description

1-Heptylamine has been used as an internal standard in the determination of biogenic monoamines and biogenic diamines by the fluorescence-HPLC method. It has also been used to study the effect of hydration on the gaseous structure of protonated heptylamine by infrared photo dissociation (IRPD) spectroscopy and computational chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Heptylamine has been used as an internal standard in the determination of biogenic monoamines and biogenic diamines by the fluorescence-HPLC method. It has also been used to study the effect of hydration on the gaseous structure of protonated heptylamine by infrared photo dissociation (IRPD) spectroscopy and computational chemistry.

Solubility
Soluble in water 6791 mg/L @ 25°C.

Notes
Air sensitive. Store away from oxidizing agents, air, acids.
Specifications

Specifications

1-Heptylamine
-23°C
154°C to 156°C
Amine-like
1.4245
MFCD00008244
UN2734
Air Sensitive
Soluble in water 6791mg/L @ 25°C.
CCCCCCCN
115.22
115.22
111-68-2
0.775
35°C (95°F)
C7H17N
CH3(CH2)6NH2
250 g
1731688
heptylamine, 1-aminoheptane, 1-heptanamine, n-heptylamine, 1-heptylamine, heptanamine, heptyl-amine, heptyl amine, dsstox_cid_681
WJYIASZWHGOTOU-UHFFFAOYSA-N
heptan-1-amine
8127
≥98%

Chemical Identifiers

111-68-2
115.22
WJYIASZWHGOTOU-UHFFFAOYSA-N
8127
CCCCCCCN
C7H17N
MFCD00008244
heptylamine, 1-aminoheptane, 1-heptanamine, n-heptylamine, 1-heptylamine, heptanamine, heptyl-amine, heptyl amine, dsstox_cid_681
heptan-1-amine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.

P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H226-H314

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, FLAMMABLE, N.O.S.

EINECSNumber : 203-895-8

RTECSNumber : MK0600000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only